Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
ENST00000419862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
ENST00000419862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
ENST00000419862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
ENST00000419862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
ENST00000419862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041654.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041654.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041654.hg.1 |
TC02002602.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0200211171.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200211171.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200211171.hg.1 |
TC0200015187.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200211171.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200211171.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
ENST00000295131.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
ENST00000419862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
ENST00000419862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
ENST00000419862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
ENST00000419862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
ENST00000419862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015187.hg.1 |
NM_182521.2 |
Refseq |