Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017315.hg.1 |
ENST00000299290.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017315.hg.1 |
ENST00000456372.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017315.hg.1 |
ENST00000536955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017315.hg.1 |
ENST00000545575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017315.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017315.hg.1 |
TC10001367.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000394982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000394982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000394982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000394982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000394982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174398.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174398.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174398.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174398.hg.1 |
TC1000010926.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000174398.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174398.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174398.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000299290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000394982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000394982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000394982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000394982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000394982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
ENST00000456372.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010926.hg.1 |
NM_152710.3 |
Refseq |