Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000074.hg.1 |
ENST00000287364.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000074.hg.1 |
ENST00000382329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000074.hg.1 |
ENST00000382331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000074.hg.1 |
ENST00000432829.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000074.hg.1 |
ENST00000453981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000074.hg.1 |
ENST00000469391.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000074.hg.1 |
NM_001190458.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000074.hg.1 |
NM_001193536.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000074.hg.1 |
NM_203447.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000074.hg.1 |
TC09000004.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0900144169.hg.1 |
ENST00000382329.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144169.hg.1 |
ENST00000382331.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144169.hg.1 |
ENST00000432829.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144169.hg.1 |
ENST00000453981.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144169.hg.1 |
ENST00000469391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144169.hg.1 |
ENST00000495184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144169.hg.1 |
NM_001190458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900144169.hg.1 |
NM_001193536.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900144169.hg.1 |
NM_203447.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900144169.hg.1 |
TC0900006438.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900144169.hg.1 |
ENST00000382329.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144169.hg.1 |
ENST00000382331.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144169.hg.1 |
ENST00000432829.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144169.hg.1 |
ENST00000453981.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144169.hg.1 |
ENST00000469391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144169.hg.1 |
ENST00000495184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144169.hg.1 |
NM_001190458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900144169.hg.1 |
NM_001193536.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900144169.hg.1 |
NM_203447.3 |
Refseq |