Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00416473_m1
          See other LYPD8 GE Assays ›
          Gene Symbol:
          LYPD8
          Gene Name:
          LY6/PLAUR domain containing 8
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 248739415 - 248755787 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00416473_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LYPD8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC033943.1 - 2 - 3 189 91

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008074 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008017 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008045 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008031 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00932702 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00932714 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008047 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008032 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008025 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008039 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21007998 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00932713 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008000 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00932701 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008048 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008051 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008046 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008034 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008026 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008023 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008021 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008018 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008040 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008062 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21007997 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008041 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008024 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00932712 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008019 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00932711 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008033 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008022 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008043 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008056 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008002 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008063 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008044 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008052 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008005 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008006 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008030 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008049 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008004 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008013 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008036 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21007999 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008038 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21008016 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 111806 AK127189.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 111806 BC031585.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 111806 BC047637.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 111806 BX649039.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 111806 NM_152389.2 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CGH Microarray 4x180K 111806 NM_194302.2 refseq_mrna_human_NM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 234377 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 492667 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 120403 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 34104 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P164998 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P164998 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 41217 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 7728 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P164998 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P164998 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008062 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008078 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008041 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008056 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008052 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008073 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008042 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008077 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008060 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008049 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008051 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008044 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008057 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008055 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008061 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008074 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008075 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008048 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008046 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008063 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008041 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008046 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008051 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008052 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008049 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008077 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008062 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008078 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008048 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008056 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008074 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008057 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008075 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008044 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008063 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008042 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008060 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008061 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008055 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21008073 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P164998 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P35545672 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P164998 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P35545672 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P10593733 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15524612 - -
          Agilent Human 1B Microarray G4111A A_32_P356227 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P35545672 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P10593733 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 238143_PM_at - -
          Affymetrix Human Genome U219 Array 11729538_PM_at - -
          Affymetrix Human Genome U219 Array 11729538_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0100018571.hg.1 ENST00000425108.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018571.hg.1 ENST00000436484.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018571.hg.1 ENST00000436484.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018571.hg.1 ENST00000436484.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018571.hg.1 ENST00000436484.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018571.hg.1 ENST00000425108.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018571.hg.1 ENST00000436484.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018571.hg.1 ENST00000436484.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018571.hg.1 ENST00000436484.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018571.hg.1 ENST00000436484.1 Ensembl
          Human Genome U133 Plus 2.0 Array 1568933_at - -
          Human Genome U133 Plus 2.0 Array 238143_PM_at - -
          Human Genome U133 Plus 2.0 Array 1568933_PM_at - -
          Human Genome U133 Plus 2.0 Array 238143_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238143_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238143_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238143_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238143_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238143_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238143_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568933_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568933_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238143_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238143_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568933_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568933_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568933_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238143_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568933_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568933_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238143_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568933_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238143_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568933_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568933_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568933_at2 - -
          Human Genome U95 Set 82950_PM_at - -
          Human Genome U95 Set 82950_at - -
          Human Genome U133 Set 238143_PM_at - -
          Human Genome U133 Set 238143_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A013795 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A013795 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 126221 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1680753 - -
          HumanHT-12 v4 Expression BeadChip ILMN_1773455 BC033943.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1773455 NM_001085474.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1773455 AY358469.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1680753 - -
          HumanHT-12 V4.0 expression beadchip ILMN_1773455 BC033943.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1773455 NM_001085474.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1773455 AY358469.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1680753 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1680753 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1680753 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1773455 BC033943.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1773455 NM_001085474.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1773455 AY358469.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1773455 BC033943.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1773455 NM_001085474.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1773455 AY358469.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix