Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007858.hg.1 |
ENST00000566439.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007858.hg.1 |
ENST00000566439.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007858.hg.1 |
ENST00000566439.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007858.hg.1 |
ENST00000566439.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007858.hg.1 |
ENST00000568711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007858.hg.1 |
ENST00000568711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007858.hg.1 |
NR_104655.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007858.hg.1 |
NR_104655.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007858.hg.1 |
NR_104655.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007858.hg.1 |
NR_104655.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007858.hg.1 |
ENST00000566439.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007858.hg.1 |
ENST00000566439.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007858.hg.1 |
ENST00000566439.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007858.hg.1 |
ENST00000566439.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007858.hg.1 |
ENST00000568711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007858.hg.1 |
ENST00000568711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007858.hg.1 |
NR_104655.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007858.hg.1 |
NR_104655.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007858.hg.1 |
NR_104655.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007858.hg.1 |
NR_104655.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157234.hg.1 |
ENST00000566439.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157234.hg.1 |
NR_104655.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157234.hg.1 |
TC1600007858.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600157234.hg.1 |
NR_104655.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157234.hg.1 |
ENST00000566439.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157235.hg.1 |
ENST00000566439.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157235.hg.1 |
NR_104655.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157235.hg.1 |
TC1600007858.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600157235.hg.1 |
ENST00000566439.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157235.hg.1 |
NR_104655.1 |
Refseq |