Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00542176_s1
          See other BRWD1-IT2 GE Assays ›
          Gene Symbol:
          BRWD1-IT2
          Gene Name:
          BRWD1 intronic transcript 2 (non-protein coding)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.21: 39313907 - 39314915 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00542176_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BRWD1-IT2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_111942.1 1 213 112
          GenBank mRNA HY025107.1 - 1 215 112

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Oligo Microarray 4x44K 23249 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 30600 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P372368 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P372368 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 177088 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P132704 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8070388 - -
          Affymetrix Human Gene 1.0 ST Array 8070387 - -
          Affymetrix Human Gene 1.1 ST Array 8070386 - -
          Affymetrix HT HG-U133+ PM Array Plate 1553069_PM_at - -
          Human Genome U133 Plus 2.0 Array 1553069_PM_at - -
          Human Genome U133 Plus 2.0 Array 1553069_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553069_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553069_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553069_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553069_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553069_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553069_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553069_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553069_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553069_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553069_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553069_at7 - -
          Human Genome U95 Set 85686_PM_at - -
          Human Genome U95 Set 85686_at - -
          Human Genome U133 Set 239602_PM_at - -
          Human Genome U133 Set 239602_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A018890 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A018890 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1683036 AF426265.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1683036 BC130301.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1683036 BC130303.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1683036 BM553164.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1683036 NR_033800.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1683036 AF426265.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1683036 BC130301.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1683036 BC130303.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1683036 BM553164.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1683036 NR_033800.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2202320 AF426265.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2202320 BC130301.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2202320 BC130303.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2202320 BM553164.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2202320 NR_033800.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1683036 AF426265.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1683036 BC130301.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1683036 BC130303.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1683036 BM553164.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1683036 NR_033800.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1683036 AF426265.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1683036 BC130301.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1683036 BC130303.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1683036 BM553164.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1683036 NR_033800.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2202320 AF426265.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2202320 BC130301.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2202320 BC130303.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2202320 BM553164.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2202320 NR_033800.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2202320 AF426265.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2202320 BC130301.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2202320 BC130303.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2202320 BM553164.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2202320 NR_033800.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1683036 AF426265.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1683036 BC130301.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1683036 BC130303.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1683036 BM553164.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1683036 NR_033800.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1683036 AF426265.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1683036 BC130301.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1683036 BC130303.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1683036 BM553164.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1683036 NR_033800.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2202320 AF426265.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2202320 BC130301.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2202320 BC130303.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2202320 BM553164.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2202320 NR_033800.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1683036 AF426265.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1683036 BC130301.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1683036 BC130303.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1683036 BM553164.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1683036 NR_033800.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2202320 AF426265.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2202320 BC130301.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2202320 BC130303.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2202320 BM553164.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2202320 NR_033800.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0