Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006109.hg.1 |
TC21000506.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006109.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006109.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006108.hg.1 |
TC21000506.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006108.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006108.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008297.hg.1 |
ENST00000478426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008297.hg.1 |
ENST00000478426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008297.hg.1 |
ENST00000478426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008297.hg.1 |
ENST00000478426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
ENST00000624077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
ENST00000624077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
ENST00000624077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
ENST00000624077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100144136.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100144136.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100144136.hg.1 |
TC2100006442.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100144136.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100144136.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008297.hg.1 |
ENST00000270162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008297.hg.1 |
ENST00000478426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008297.hg.1 |
ENST00000478426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008297.hg.1 |
ENST00000478426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008297.hg.1 |
ENST00000478426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
ENST00000624077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
ENST00000624077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
ENST00000624077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
ENST00000624077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006442.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100144135.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100144135.hg.1 |
NM_173354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100144135.hg.1 |
TC2100006442.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100144135.hg.1 |
ENST00000613488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100144135.hg.1 |
NM_173354.4 |
Refseq |