Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00545030_g1
          See other SIK1 GE Assays ›
          Gene Symbol:
          SIK1 LOC102724428
          Gene Name:
          salt inducible kinase 1
          serine/threonine-protein kinase SIK1
          Gene Aliases:
          EIEE30, MSK, SIK, SNF1LK
          -
          Chromosome Location:
          Chr.21: 43414490 - 43427126 on Build GRCh38
          Chromosome Location:
          Chr.21: 6111134 - 6123778 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs00545030_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SIK1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173354.3 NP_775490.2 13 - 14 2104 55
          XM_011529474.2 XP_011527776.1 12 - 13 1961 55
          GenBank mRNA AB047786.1 - 13 - 14 2104 55
          Gene Symbol: LOC102724428
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001320643.1 NP_001307572.1 13 - 14 2110 55
          XM_017028208.1 XP_016883697.1 12 - 13 1962 55
          GenBank mRNA AK131076.1 - 12 - 13 1921 55
          BC038504.1 - 13 - 14 2110 4831049 55

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2100006442.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC21000506.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SIK1
          Click here to find siRNAs for Gene LOC102724428

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of sodium ion transport
          protein phosphorylation
          cell cycle
          regulation of mitotic cell cycle
          regulation of myotube differentiation
          negative regulation of triglyceride biosynthetic process
          negative regulation of CREB transcription factor activity
          cellular response to hormone stimulus
          intracellular signal transduction
          entrainment of circadian clock by photoperiod
          regulation of cell differentiation
          negative regulation of gluconeogenesis
          protein autophosphorylation
          rhythmic process
          cardiac muscle cell differentiation
          positive regulation of anoikis
          magnesium ion binding
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          cAMP response element binding protein binding
          protein kinase binding
          14-3-3 protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008297.hg.1 ENST00000478426.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008297.hg.1 ENST00000478426.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008297.hg.1 ENST00000478426.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008297.hg.1 ENST00000478426.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21006105.hg.1 TC21000506.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR21006105.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21006105.hg.1 NM_173354.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 ENST00000624077.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 ENST00000624077.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 ENST00000624077.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 ENST00000624077.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 ENST00000624077.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 ENST00000624077.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 ENST00000624077.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 ENST00000624077.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Clariom D Assay TC2100006442.hg.1 NM_173354.4 Refseq
          Affymetrix Human Clariom D Assay TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Clariom D Assay TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Clariom D Assay TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Clariom D Assay TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Clariom D Assay TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Clariom D Assay TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Clariom D Assay TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Clariom D Assay TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Clariom D Assay TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Clariom D Assay TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Clariom D Assay TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Clariom D Assay TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Clariom D Assay TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Clariom D Assay TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Clariom D Assay TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Clariom D Assay TC2100008297.hg.1 ENST00000270162.6 Ensembl
          Affymetrix Human Clariom D Assay TC2100008297.hg.1 ENST00000478426.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100008297.hg.1 ENST00000478426.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100008297.hg.1 ENST00000478426.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100008297.hg.1 ENST00000478426.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100144139.hg.1 ENST00000613488.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2100144139.hg.1 NM_173354.4 Refseq
          Affymetrix Human Clariom D Assay PSR2100144139.hg.1 TC2100006442.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100144139.hg.1 NM_173354.4 Refseq
          Affymetrix Human Clariom D Assay PSR2100144139.hg.1 ENST00000613488.2 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0