Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011191.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011191.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011191.hg.1 |
TC09000846.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0900163672.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163672.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163672.hg.1 |
TC0900009276.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900163672.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163672.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
ENST00000343053.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009276.hg.1 |
NM_015456.4 |
Refseq |