Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00609977_m1
          See other NGFR GE Assays ›
          Gene Symbol:
          NGFR
          Gene Name:
          nerve growth factor receptor
          Gene Aliases:
          CD271, Gp80-LNGFR, TNFRSF16, p75(NTR), p75NTR
          Chromosome Location:
          Chr.17: 49495293 - 49515020 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00609977_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NGFR
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002507.3 NP_002498.1 3 - 4 689 140
          GenBank mRNA AK303278.1 - 3 - 4 757 140
          AK313654.1 - 3 - 4 689 140
          BC050309.1 - 3 - 4 689 5263715 140
          M14764.1 - 3 - 4 677 140

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700008209.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17000645.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NGFR

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          inflammatory response
          immune response
          axon guidance
          response to wounding
          negative regulation of neuron projection development
          sensory perception of pain
          membrane protein intracellular domain proteolysis
          positive regulation of myelination
          positive regulation of synaptic transmission, cholinergic
          response to lipopolysaccharide
          circadian regulation of gene expression
          tumor necrosis factor-mediated signaling pathway
          cellular response to oxidative stress
          positive regulation of Rho protein signal transduction
          skeletal muscle cell differentiation
          regulation of cell population proliferation
          glucose homeostasis
          regulation of apoptotic process
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of MAPK cascade
          negative regulation of neuron apoptotic process
          neurotrophin TRK receptor signaling pathway
          negative regulation of axonogenesis
          positive regulation of axonogenesis
          positive regulation of protein kinase B signaling
          negative regulation of mitochondrial depolarization
          positive regulation of synaptic transmission, glutamatergic
          apoptotic signaling pathway
          positive regulation of neuron death
          regulation of reactive oxygen species metabolic process
          amyloid-beta binding
          transmembrane signaling receptor activity
          tumor necrosis factor-activated receptor activity
          neurotrophin TRKA receptor binding
          protein binding
          ubiquitin protein ligase binding
          signaling receptor activity
          nerve growth factor binding
          preprotein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR17008028.hg.1 ENST00000172229.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17008028.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17008028.hg.1 NM_002507.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17008028.hg.1 TC17000645.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17008027.hg.1 ENST00000172229.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17008027.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17008027.hg.1 NM_002507.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17008027.hg.1 TC17000645.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000509200.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000509200.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000509200.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Clariom D Assay PSR1700160713.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1700160713.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700160713.hg.1 NM_002507.3 Refseq
          Affymetrix Human Clariom D Assay PSR1700160713.hg.1 TC1700008209.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700160713.hg.1 NM_002507.3 Refseq
          Affymetrix Human Clariom D Assay PSR1700160713.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1700160713.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 ENST00000509200.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 ENST00000509200.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 ENST00000509200.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Clariom D Assay TC1700008209.hg.1 NM_002507.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0