Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008747.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008747.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008747.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008747.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008747.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008747.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008747.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008747.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008747.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008747.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008747.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008747.hg.1 |
NM_018558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008747.hg.1 |
NM_018558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008747.hg.1 |
NM_018558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008747.hg.1 |
NM_018558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008747.hg.1 |
NM_018558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008747.hg.1 |
NM_018558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008747.hg.1 |
NM_018558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008747.hg.1 |
NM_018558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008747.hg.1 |
NM_018558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008747.hg.1 |
NM_018558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009513.hg.1 |
ENST00000370306.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009513.hg.1 |
NM_018558.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009513.hg.1 |
TC0X000738.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0X00159477.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159477.hg.1 |
NM_018558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159477.hg.1 |
TC0X00008747.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00159477.hg.1 |
NM_018558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159477.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008747.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008747.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008747.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008747.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008747.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008747.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008747.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008747.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008747.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008747.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008747.hg.1 |
ENST00000598523.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008747.hg.1 |
NM_018558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008747.hg.1 |
NM_018558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008747.hg.1 |
NM_018558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008747.hg.1 |
NM_018558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008747.hg.1 |
NM_018558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008747.hg.1 |
NM_018558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008747.hg.1 |
NM_018558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008747.hg.1 |
NM_018558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008747.hg.1 |
NM_018558.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008747.hg.1 |
NM_018558.3 |
Refseq |