Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00697000_g1
          See other BRAF GE Assays ›
          Gene Symbol:
          BRAF KIAA1549
          Gene Name:
          B-Raf proto-oncogene, serine/threonine kinase
          KIAA1549
          Gene Aliases:
          B-RAF1, B-raf, BRAF1, NS7, RAFB1
          -
          Chromosome Location:
          Chr.7: 140719331 - 140924764 on Build GRCh38
          Chromosome Location:
          Chr.7: 138831377 - 138981626 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs00697000_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BRAF
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004333.4 NP_004324.2 10 - 11 1371 77
          XM_005250045.2 XP_005250102.1 10 - 11 1318 77
          XM_017012558.1 XP_016868047.1 11 - 12 1438 77
          XM_017012559.1 XP_016868048.1 11 - 12 1438 77
          GenBank mRNA AK314346.1 - 10 - 11 1318 77
          BC101757.1 - 10 - 11 1332 8069263 77
          BC112079.1 - 10 - 11 1332 8327547 77
          HM437233.1 - 2 - 3 171 77
          M95712.2 - 10 - 11 1371 77
          Gene Symbol: KIAA1549
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AM989477.1 - 1517 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700013629.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07001927.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene BRAF
          Click here to find siRNAs for Gene KIAA1549

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          myeloid progenitor cell differentiation
          protein phosphorylation
          visual learning
          animal organ morphogenesis
          positive regulation of gene expression
          negative regulation of fibroblast migration
          thyroid gland development
          positive regulation of peptidyl-serine phosphorylation
          somatic stem cell population maintenance
          regulation of cell population proliferation
          negative regulation of apoptotic process
          CD4-positive, alpha-beta T cell differentiation
          positive T cell selection
          response to peptide hormone
          negative regulation of neuron apoptotic process
          thymus development
          positive regulation of axon regeneration
          positive regulation of axonogenesis
          protein heterooligomerization
          positive regulation of stress fiber assembly
          response to cAMP
          long-term synaptic potentiation
          head morphogenesis
          face development
          positive regulation of ERK1 and ERK2 cascade
          cellular response to calcium ion
          cellular response to xenobiotic stimulus
          establishment of protein localization to membrane
          positive regulation of substrate adhesion-dependent cell spreading
          glucose transmembrane transport
          negative regulation of synaptic vesicle exocytosis
          negative regulation of endothelial cell apoptotic process
          protein kinase activity
          protein serine/threonine kinase activity
          MAP kinase kinase kinase activity
          calcium ion binding
          protein binding
          ATP binding
          small GTPase binding
          mitogen-activated protein kinase kinase binding
          identical protein binding
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000469930.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000469930.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000469930.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000469930.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000469930.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07029949.hg.1 ENST00000288602.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07029949.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07029949.hg.1 TC07001927.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000469930.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000469930.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000469930.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000469930.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000469930.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay PSR0700202207.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0700202207.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700202207.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay PSR0700202207.hg.1 TC0700013629.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700202207.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0700202207.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700202207.hg.1 NM_004333.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0