Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00892966_g1
          See other PTPRF GE Assays ›
          Gene Symbol:
          PTPRF
          Gene Name:
          protein tyrosine phosphatase, receptor type F
          Gene Aliases:
          BNAH2, LAR
          Chromosome Location:
          Chr.1: 43522238 - 43623672 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00892966_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PTPRF
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_005271079.3 XP_005271136.1 18 - 19 3495 77
          XM_005271080.3 XP_005271137.1 17 - 18 3483 77
          XM_005271081.3 XP_005271138.1 16 - 17 3456 77
          XM_005271082.3 XP_005271139.1 15 - 16 3150 77
          XM_006710795.3 XP_006710858.1 19 - 20 3473 77
          XM_006710796.3 XP_006710859.1 17 - 18 3468 77
          XM_006710797.3 XP_006710860.1 18 - 19 3495 77
          XM_006710798.3 XP_006710861.1 17 - 18 3468 77
          XM_006710799.3 XP_006710862.1 17 - 18 3189 77
          XM_006710800.3 XP_006710863.1 16 - 17 3162 77
          XM_006710801.3 XP_006710864.1 17 - 18 2916 77
          XM_011541871.2 XP_011540173.1 19 - 20 3507 77
          XM_011541872.2 XP_011540174.1 19 - 20 3493 77
          XM_011541873.2 XP_011540175.1 19 - 20 3427 77
          XM_017001942.1 XP_016857431.1 16 - 17 3124 77
          XM_017001943.1 XP_016857432.1 16 - 17 3177 77
          XM_017001944.1 XP_016857433.1 16 - 17 3162 77
          XM_017001945.1 XP_016857434.1 15 - 16 3150 77
          XM_017001946.1 XP_016857435.1 16 - 17 2610 77
          XM_017001947.1 XP_016857436.1 15 - 16 2583 77
          XM_017001948.1 XP_016857437.1 15 - 16 2609 77
          XM_017001949.1 XP_016857438.1 16 - 17 2574 77
          XM_017001950.1 XP_016857439.1 14 - 15 2579 77
          GenBank mRNA AB177856.1 - 16 - 17 2107 77
          AB177857.1 - 16 - 17 3415 77
          AB209478.1 - 8 - 9 2064 77
          AK125325.1 - 7 - 8 1367 77
          AK127007.1 - 4 - 5 310 77
          BC048768.1 - 16 - 17 3426 5502598 77
          BX537361.1 - 4 - 5 1627 77
          Y00815.1 - 17 - 18 3467 77

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          transmembrane receptor protein tyrosine phosphatase signaling pathway
          chemical synaptic transmission
          cell migration
          neuron projection regeneration
          peptidyl-tyrosine dephosphorylation
          regulation of axon regeneration
          negative regulation of receptor binding
          protein tyrosine phosphatase activity
          transmembrane receptor protein tyrosine phosphatase activity
          heparin binding
          chondroitin sulfate proteoglycan binding
          protein-containing complex binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0