Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030979.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030979.hg.1 |
ENST00000485972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030979.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030979.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030979.hg.1 |
TC07002027.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000469108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000469108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000469108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000469108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000469108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000469108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000469108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000469108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000476691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000476691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000476691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000476691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000487703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000487703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000487703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000487703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000487703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000487703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000487703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
ENST00000487703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000469108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000469108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000469108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000469108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000469108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000469108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000469108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000469108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000476691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000476691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000476691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000476691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000487703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000487703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000487703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000487703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000487703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000487703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000487703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
ENST00000487703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013055.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194091.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194091.hg.1 |
ENST00000469108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194091.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194091.hg.1 |
ENST00000487703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194091.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194091.hg.1 |
NM_004935.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194091.hg.1 |
TC0700013055.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700194091.hg.1 |
ENST00000485972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194091.hg.1 |
ENST00000297518.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194091.hg.1 |
ENST00000469108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194091.hg.1 |
ENST00000487703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194091.hg.1 |
NM_001164410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194091.hg.1 |
NM_004935.3 |
Refseq |