Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00895891_m1
          See other FUBP3 GE Assays ›
          Gene Symbol:
          FUBP3
          Gene Name:
          far upstream element binding protein 3
          Gene Aliases:
          FBP3
          Chromosome Location:
          Chr.9: 130579567 - 130638352 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00895891_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FUBP3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003934.1 NP_003925.1 12 - 13 1222 68
          XM_005272232.1 XP_005272289.1 12 - 13 1228 68
          XM_006717313.2 XP_006717376.1 13 - 14 2073 68
          XM_011519172.2 XP_011517474.1 12 - 13 1228 68
          XM_011519173.2 XP_011517475.1 10 - 11 1480 68
          XM_011519174.2 XP_011517476.1 14 - 15 2318 68
          XM_017015269.1 XP_016870758.1 13 - 14 2073 68
          XM_017015270.1 XP_016870759.1 12 - 13 2015 68
          XM_017015271.1 XP_016870760.1 13 - 14 2260 68
          XM_017015272.1 XP_016870761.1 10 - 11 1043 68
          XM_017015273.1 XP_016870762.1 12 - 13 2015 68
          GenBank mRNA AK295155.1 - 12 - 13 1222 68
          BC001325.1 - 4 - 5 382 68
          BC137338.1 - 12 - 13 1219 9021336 68
          BC137340.1 - 12 - 13 1219 9021338 68
          U69127.1 - 12 - 13 1198 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900008959.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09000738.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          transcription by RNA polymerase II
          positive regulation of gene expression
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          RNA binding
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000465949.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000465949.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000465949.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000465949.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000465949.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000467100.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000467100.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000467100.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000467100.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000467100.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000467100.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000467100.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000472006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000472006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000472006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000472006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000487406.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000487406.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000487406.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000487406.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000487406.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000487406.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000487406.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000487406.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000492199.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000492199.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000492199.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 ENST00000492199.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09009345.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09009345.hg.1 NM_003934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09009345.hg.1 TC09000738.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000465949.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000465949.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000465949.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000465949.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000465949.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000467100.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000467100.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000467100.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000467100.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000467100.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000467100.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000467100.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000472006.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000472006.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000472006.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000472006.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000487406.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000487406.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000487406.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000487406.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000487406.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000487406.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000487406.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000487406.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000492199.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000492199.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000492199.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 ENST00000492199.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008959.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900160912.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0900160912.hg.1 ENST00000487406.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900160912.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900160912.hg.1 TC0900008959.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900160912.hg.1 NM_003934.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900160912.hg.1 ENST00000319725.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0900160912.hg.1 ENST00000487406.2 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix