Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00896290_m1
          See other PROX1 GE Assays ›
          Gene Symbol:
          PROX1
          Gene Name:
          prospero homeobox 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 213983239 - 214041510 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00896290_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PROX1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_011509773.2 XP_011508075.1 1 - 2 89 100

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_011509773.2 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          cell fate determination
          kidney development
          liver development
          positive regulation of endothelial cell proliferation
          lymphangiogenesis
          lens development in camera-type eye
          hepatocyte cell migration
          transcription, DNA-templated
          brain development
          circadian rhythm
          positive regulation of cell population proliferation
          negative regulation of cell population proliferation
          regulation of gene expression
          positive regulation of endothelial cell migration
          dorsal spinal cord development
          dentate gyrus development
          cerebellar granule cell differentiation
          neural tube development
          neuron differentiation
          skeletal muscle thin filament assembly
          lung development
          olfactory placode formation
          pancreas development
          response to nutrient levels
          regulation of circadian rhythm
          otic placode formation
          negative regulation of DNA-binding transcription factor activity
          negative regulation of viral genome replication
          positive regulation of cyclin-dependent protein serine/threonine kinase activity
          positive regulation of cell cycle
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          lens placode formation involved in camera-type eye formation
          venous blood vessel morphogenesis
          ventricular cardiac myofibril assembly
          cardiac muscle cell differentiation
          atrial cardiac muscle tissue morphogenesis
          ventricular cardiac muscle tissue morphogenesis
          retina morphogenesis in camera-type eye
          embryonic retina morphogenesis in camera-type eye
          endocardium formation
          positive regulation of sarcomere organization
          ventricular septum morphogenesis
          aorta smooth muscle tissue morphogenesis
          positive regulation of heart growth
          lymphatic endothelial cell differentiation
          branching involved in pancreas morphogenesis
          lens fiber cell morphogenesis
          hepatocyte differentiation
          negative regulation of bile acid biosynthetic process
          hepatocyte proliferation
          acinar cell differentiation
          neuronal stem cell population maintenance
          positive regulation of cell cycle checkpoint
          positive regulation of neural precursor cell proliferation
          positive regulation of forebrain neuron differentiation
          transcription cis-regulatory region binding
          DNA-binding transcription factor activity, RNA polymerase II-specific
          core promoter sequence-specific DNA binding
          DNA binding
          DNA-binding transcription factor activity
          transcription corepressor activity
          protein binding
          nuclear receptor binding
          DNA binding domain binding
          LBD domain binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000498508.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000498508.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000498508.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000498508.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000607425.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000607425.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000607425.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000607726.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000607726.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_001270616.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_001270616.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_001270616.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_001270616.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_001270616.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_001270616.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_001270616.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_001270616.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_001270616.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_001270616.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_001270616.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000366958.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000366958.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000366958.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000366958.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000366958.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000366958.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000366958.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000366958.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000366958.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000366958.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000435016.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000435016.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000435016.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000435016.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000435016.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000435016.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000435016.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000435016.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000435016.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000435016.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000435016.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000471129.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000471129.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000471129.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000471129.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000498508.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000498508.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000498508.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000498508.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000498508.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000498508.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011566.hg.1 ENST00000498508.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000498508.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000498508.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000498508.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000498508.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000607425.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000607425.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000607425.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000607726.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000607726.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_001270616.1 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_001270616.1 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_001270616.1 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_001270616.1 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_001270616.1 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_001270616.1 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_001270616.1 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_001270616.1 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_001270616.1 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_001270616.1 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_001270616.1 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 NM_002763.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000261454.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000366958.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000366958.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000366958.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000366958.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000366958.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000366958.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000366958.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000366958.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000366958.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000366958.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000435016.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000435016.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000435016.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000435016.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000435016.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000435016.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000435016.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000435016.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000435016.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000435016.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000435016.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000471129.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000471129.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000471129.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000471129.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000498508.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000498508.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000498508.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000498508.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000498508.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000498508.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011566.hg.1 ENST00000498508.6 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0