Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00896500_s1
          See other PRKAG2-AS1 GE Assays ›
          Gene Symbol:
          PRKAG2-AS1
          Gene Name:
          PRKAG2 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 151877042 - 151879223 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00896500_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PRKAG2-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038926.1 3 475 160
          GenBank mRNA BC031308.1 - 1 352 160
          CA419137.1 - 1 160
          KR153483.1 - 2 746 160
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BE669437.1 - 1 Chr. 7 - 151878715 - 151879214 [-] on Build GRCh38 160
          GenBankmRNA BE220250.1 - 1 Chr. 7 - 151878788 - 151879214 [-] on Build GRCh38 160
          GenBankmRNA BE348405.1 - 1 Chr. 7 - 151878777 - 151879212 [-] on Build GRCh38 160
          GenBankmRNA AI912720.1 - 1 Chr. 7 - 151878690 - 151879212 [-] on Build GRCh38 160
          GenBankmRNA AW136482.1 - 1 Chr. 7 - 151878750 - 151879214 [-] on Build GRCh38 160
          GenBankmRNA BU596754.1 - 2 494 Chr. 7 - 151877721 - 151879208 [+] on Build GRCh38 160
          GenBankmRNA CK904908.1 - 1 167 Chr. 7 - 151878741 - 151879214 [+] on Build GRCh38 160
          GenBankmRNA CK904907.1 - 1 Chr. 7 - 151878740 - 151879214 [-] on Build GRCh38 160
          GenBankmRNA AW970759.1 - 1 Chr. 7 - 151878734 - 151879212 [-] on Build GRCh38 160
          GenBankmRNA DB351264.1 - 1 Chr. 7 - 151878663 - 151879213 [-] on Build GRCh38 160
          GenBankmRNA BI551278.1 - 1 352 Chr. 7 - 151878634 - 151879214 [+] on Build GRCh38 160
          GenBankmRNA AA535276.1 - 1 Chr. 7 - 151878745 - 151879214 [-] on Build GRCh38 160
          GenBankmRNA AA775841.1 - 2 378 Chr. 7 - 151877834 - 151878997 [+] on Build GRCh38 160
          GenBankmRNA BG924694.1 - 2 463 Chr. 7 - 151877839 - 151879122 [+] on Build GRCh38 160
          GenBankmRNA AI949265.1 - 1 Chr. 7 - 151878686 - 151879214 [-] on Build GRCh38 160

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream protein kinase AMP-activated non-catalytic subunit gamma 2 PRKAG2 AAKG,
          AAKG2,
          CMH6,
          H91620p,
          WPWS
          -1.5kb Sense Chr. 7 - 151556114 - 151877231 on NCBI Build GRCh38
          Downstream polypeptide N-acetylgalactosaminyltransferase-like 5 GALNTL5 GALNACT19,
          GALNT15,
          GalNAc-T5L
          77.2kb Anti-sense Chr. 7 - 151956379 - 152019934 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700009719.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07001020.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05874142 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05874143 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05874144 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P735306 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 12947 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P735306 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P38256646 - -
          Agilent Human 1B Microarray G4111A A_32_P181391 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR07015823.hg.1 ENST00000464464.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07015823.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07015823.hg.1 NR_038926.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07015823.hg.1 TC07001020.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07015822.hg.1 ENST00000464464.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07015822.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07015822.hg.1 NR_038926.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07015822.hg.1 TC07001020.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07015824.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07015824.hg.1 NR_038926.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07015824.hg.1 TC07001020.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07015820.hg.1 ENST00000464464.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07015820.hg.1 NR_038926.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07015820.hg.1 TC07001020.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 ENST00000464464.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 ENST00000464464.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 ENST00000464464.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 ENST00000464464.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 NR_038926.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 NR_038926.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 NR_038926.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 NR_038926.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 NR_038926.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 NR_038926.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 NR_038926.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009719.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700169385.hg.1 ENST00000464464.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169385.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169385.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169385.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700169385.hg.1 TC0700009719.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700169385.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700169385.hg.1 ENST00000464464.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169385.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169385.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169387.hg.1 ENST00000464464.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169387.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169387.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169387.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700169387.hg.1 TC0700009719.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700169387.hg.1 ENST00000464464.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169387.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169387.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169387.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700169388.hg.1 ENST00000464464.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169388.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169388.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169388.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700169388.hg.1 TC0700009719.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700169388.hg.1 ENST00000464464.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169388.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169388.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169388.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700169391.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169391.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169391.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700169391.hg.1 TC0700009719.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700169391.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169391.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169391.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700169377.hg.1 ENST00000464464.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169377.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700169377.hg.1 TC0700009719.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700169377.hg.1 ENST00000464464.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169377.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700169389.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169389.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169389.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700169389.hg.1 TC0700009719.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700169389.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169389.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169389.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700169390.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169390.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169390.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700169390.hg.1 TC0700009719.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700169390.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169390.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169390.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700169394.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169394.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169394.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700169394.hg.1 TC0700009719.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700169394.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169394.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169394.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 ENST00000464464.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 ENST00000464464.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 ENST00000464464.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 ENST00000464464.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 ENST00000467458.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 NR_038926.1 Refseq
          Affymetrix Human Clariom D Assay TC0700009719.hg.1 NR_038926.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035352 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035353 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035353 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035352 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 232317 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0