Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010427.hg.1 |
TC20000780.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010427.hg.1 |
ENST00000217426.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010427.hg.1 |
ENST00000468908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010427.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010427.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010427.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000468908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000468908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000468908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000468908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000468908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000468908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000468908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000473516.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000473516.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000473516.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000512005.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000512005.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000512005.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000606061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000606061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000606061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000606061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000606061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000606061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000606061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
ENST00000606061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000468908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000468908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000468908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000468908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000468908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000468908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000468908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000473516.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000473516.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000473516.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000512005.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000512005.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000512005.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000606061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000606061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000606061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000606061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000606061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000606061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000606061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
ENST00000606061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008915.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000162135.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162135.hg.1 |
ENST00000468908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162135.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162135.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162135.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000162135.hg.1 |
NM_001161766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000162135.hg.1 |
TC2000008915.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000162135.hg.1 |
ENST00000480653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162135.hg.1 |
ENST00000217426.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162135.hg.1 |
ENST00000468908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162135.hg.1 |
ENST00000538132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162135.hg.1 |
NM_000687.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000162135.hg.1 |
NM_001161766.1 |
Refseq |