Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00902318_m1
          See other MAPT GE Assays ›
          Gene Symbol:
          MAPT
          Gene Name:
          microtubule associated protein tau
          Gene Aliases:
          DDPAC, FTDP-17, MAPTL, MSTD, MTBT1, MTBT2, PPND, PPP1R103, TAU
          Chromosome Location:
          Chr.17: 45894382 - 46028334 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00902318_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MAPT
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001123066.3 NP_001116538.2 7 - 8 1510 133
          NM_016835.4 NP_058519.3 7 - 8 1510 133
          XM_005257362.4 XP_005257419.1 7 - 8 1602 133
          XM_005257364.4 XP_005257421.1 6 - 7 1515 133
          XM_005257365.4 XP_005257422.1 7 - 8 1602 133
          XM_005257366.3 XP_005257423.1 5 - 6 1470 133
          XM_005257369.4 XP_005257426.1 6 - 7 537 133
          XM_005257370.4 XP_005257427.1 5 - 6 514 133
          XM_005257371.4 XP_005257428.1 4 - 5 363 133
          GenBank mRNA AW295014.1 - 2 - 3 133

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700008082.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17000592.hg.1, TC17_ctg5_hap1000017.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MAPT

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          neuron migration
          brain development
          female pregnancy
          response to nutrient
          adult walking behavior
          axo-dendritic transport
          intrinsic apoptotic signaling pathway in response to oxidative stress
          response to organic substance
          regulation of autophagy
          regulation of microtubule polymerization
          positive regulation of microtubule polymerization
          negative regulation of intracellular transport
          positive regulation of axon extension
          mitochondrion transport along microtubule
          axon extension
          generation of neurons
          regulation of microtubule-based movement
          structural constituent of cytoskeleton
          protein binding
          microtubule binding
          SH3 domain binding
          enzyme binding
          protein kinase binding
          apolipoprotein binding
          protein-containing complex binding
          protein phosphatase 2A binding
          lipoprotein particle binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR17_ctg5_hap1000220.hg.1 NM_001123066.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17_ctg5_hap1000220.hg.1 NM_016835.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17_ctg5_hap1000220.hg.1 TC17_ctg5_hap1000017.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17007254.hg.1 ENST00000262410.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17007254.hg.1 ENST00000344290.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17007254.hg.1 ENST00000415613.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17007254.hg.1 NM_001123066.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17007254.hg.1 NM_016835.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17007254.hg.1 TC17000592.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1700159253.hg.1 ENST00000262410.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1700159253.hg.1 ENST00000344290.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1700159253.hg.1 ENST00000415613.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1700159253.hg.1 ENST00000571987.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700159253.hg.1 NM_001123066.3 Refseq
          Affymetrix Human Clariom D Assay PSR1700159253.hg.1 NM_016835.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700159253.hg.1 TC1700008082.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700159253.hg.1 ENST00000415613.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1700159253.hg.1 ENST00000262410.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1700159253.hg.1 ENST00000344290.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1700159253.hg.1 ENST00000571987.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700159253.hg.1 NM_001123066.3 Refseq
          Affymetrix Human Clariom D Assay PSR1700159253.hg.1 NM_016835.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0