Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00906335_g1
          See other OSBPL9 GE Assays ›
          Gene Symbol:
          OSBPL9
          Gene Name:
          oxysterol binding protein like 9
          Gene Aliases:
          ORP-9, ORP9
          Chromosome Location:
          Chr.1: 51616874 - 51789219 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00906335_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OSBPL9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024586.5 NP_078862.4 19 - 20 1932 138
          NM_148904.3 NP_683702.1 15 - 16 1649 138
          NM_148905.3 NP_683703.1 14 - 15 1553 138
          NM_148906.2 NP_683704.2 18 - 19 1881 138
          NM_148907.2 NP_683705.1 15 - 16 1526 138
          NM_148908.3 NP_683706.3 18 - 19 1893 138
          NM_148909.3 NP_683707.3 19 - 20 1962 138
          NR_036662.1 18 - 19 1853 138
          XM_006710318.2 XP_006710381.1 18 - 19 1692 138
          XM_006710320.3 XP_006710383.1 16 - 17 1529 138
          XM_006710321.2 XP_006710384.1 15 - 16 1525 138
          XM_006710322.3 XP_006710385.1 16 - 17 1686 138
          XM_006710323.2 XP_006710386.1 15 - 16 1594 138
          XM_006710324.3 XP_006710387.1 17 - 18 1604 138
          XM_006710325.3 XP_006710388.1 16 - 17 1574 138
          XM_006710326.2 XP_006710389.1 14 - 15 1576 138
          XM_011540599.2 XP_011538901.1 20 - 21 1786 138
          XM_011540600.2 XP_011538902.1 19 - 20 1753 138
          XM_011540601.2 XP_011538903.1 18 - 19 1714 138
          XM_011540602.2 XP_011538904.1 19 - 20 1688 138
          XM_011540603.2 XP_011538905.1 19 - 20 1712 138
          XM_011540604.2 XP_011538906.1 16 - 17 1487 138
          XM_017000218.1 XP_016855707.1 17 - 18 1653 138
          XM_017000219.1 XP_016855708.1 18 - 19 1673 138
          XM_017000220.1 XP_016855709.1 15 - 16 1414 138
          XM_017000221.1 XP_016855710.1 15 - 16 1365 138
          XM_017000222.1 XP_016855711.1 19 - 20 1898 138
          XM_017000223.1 XP_016855712.1 19 - 20 1896 138
          XM_017000224.1 XP_016855713.1 16 - 17 1841 138
          XM_017000225.1 XP_016855714.1 17 - 18 1692 138
          GenBank mRNA AF392445.1 - 19 - 20 1706 138
          AK022554.1 - 14 - 15 1333 138
          AK027535.1 - 18 - 19 1663 138
          AK027707.1 - 15 - 16 1489 138
          AK056617.1 - 19 - 20 1751 138
          AK074392.1 - 5 - 6 465 138
          AK091703.1 - 18 - 19 1635 138
          AK124037.1 - 10 - 11 2179 138
          AK128043.1 - 21 - 22 2156 138
          AK297656.1 - 16 - 17 1679 138
          AK315167.1 - 15 - 16 1522 138
          AY178997.1 - 18 - 19 1655 138
          BC025978.2 - 15 - 16 1559 3621193 138
          CR457288.1 - 12 - 13 1200 138

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100008289.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01000631.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene OSBPL9

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid transport
          lipid binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01010401.hg.1 ENST00000337809.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01010401.hg.1 ENST00000361556.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01010401.hg.1 ENST00000371710.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01010401.hg.1 ENST00000371714.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01010401.hg.1 ENST00000428468.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01010401.hg.1 ENST00000435686.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01010401.hg.1 ENST00000447887.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01010401.hg.1 ENST00000453295.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01010401.hg.1 ENST00000462759.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01010401.hg.1 ENST00000486942.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01010401.hg.1 ENST00000530544.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01010401.hg.1 ENST00000531828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01010401.hg.1 NM_024586.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01010401.hg.1 NM_148904.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01010401.hg.1 NM_148905.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01010401.hg.1 NM_148906.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01010401.hg.1 NM_148907.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01010401.hg.1 NM_148908.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01010401.hg.1 NM_148909.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01010401.hg.1 NR_036662.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01010401.hg.1 TC01000631.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000361556.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000371714.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000428468.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000435274.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000447887.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000453295.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000462759.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000475697.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000486942.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000495776.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000531819.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000531828.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 NM_024586.5 Refseq
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 NM_148904.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 NM_148905.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 NM_148906.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 NM_148907.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 NM_148908.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 NM_148909.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 NR_036662.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 TC0100008289.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000361556.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000371714.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000428468.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000435274.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000475697.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000447887.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000453295.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000462759.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000486942.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000495776.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000531819.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 ENST00000531828.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 NM_024586.5 Refseq
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 NM_148904.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 NM_148905.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 NM_148906.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 NM_148907.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 NM_148908.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 NM_148909.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100159115.hg.1 NR_036662.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0