Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013389.hg.1 |
ENST00000286317.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013389.hg.1 |
ENST00000286317.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013389.hg.1 |
ENST00000286317.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013389.hg.1 |
ENST00000286317.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013389.hg.1 |
ENST00000286317.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013389.hg.1 |
ENST00000286317.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013389.hg.1 |
ENST00000286317.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013389.hg.1 |
ENST00000420343.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013389.hg.1 |
ENST00000420343.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013389.hg.1 |
ENST00000420343.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013389.hg.1 |
ENST00000420343.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013389.hg.1 |
ENST00000524289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013389.hg.1 |
ENST00000524289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013389.hg.1 |
ENST00000524289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013389.hg.1 |
ENST00000524289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013389.hg.1 |
NM_001100816.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013389.hg.1 |
NM_001100816.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013389.hg.1 |
NM_001100816.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013389.hg.1 |
NM_001100816.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013389.hg.1 |
NM_004270.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013389.hg.1 |
NM_004270.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013389.hg.1 |
NM_004270.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013389.hg.1 |
NM_004270.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027576.hg.1 |
ENST00000420343.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027576.hg.1 |
NM_001100816.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027576.hg.1 |
TC05003435.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0500197297.hg.1 |
ENST00000420343.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500197297.hg.1 |
NM_001100816.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500197297.hg.1 |
TC0500013389.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500197297.hg.1 |
NM_001100816.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500197297.hg.1 |
ENST00000420343.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013389.hg.1 |
ENST00000286317.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013389.hg.1 |
ENST00000286317.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013389.hg.1 |
ENST00000286317.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013389.hg.1 |
ENST00000286317.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013389.hg.1 |
ENST00000286317.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013389.hg.1 |
ENST00000286317.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013389.hg.1 |
ENST00000286317.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013389.hg.1 |
ENST00000420343.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013389.hg.1 |
ENST00000420343.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013389.hg.1 |
ENST00000420343.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013389.hg.1 |
ENST00000420343.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013389.hg.1 |
ENST00000524289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013389.hg.1 |
ENST00000524289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013389.hg.1 |
ENST00000524289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013389.hg.1 |
ENST00000524289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013389.hg.1 |
NM_001100816.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013389.hg.1 |
NM_001100816.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013389.hg.1 |
NM_001100816.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013389.hg.1 |
NM_001100816.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013389.hg.1 |
NM_004270.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013389.hg.1 |
NM_004270.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013389.hg.1 |
NM_004270.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013389.hg.1 |
NM_004270.4 |
Refseq |