Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000428061.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000428061.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000428061.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000428061.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000428061.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000450866.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000610387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000610387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000610387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000610387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000610387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000610387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000610387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000610387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000610387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
ENST00000610387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_001199319.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_001199319.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_001199319.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_001199319.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_001199319.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_001199319.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_001199319.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_001199319.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_001199319.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_001199319.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009186.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22000218.hg.1 |
TC22001451.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22000218.hg.1 |
ENST00000329627.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22000218.hg.1 |
ENST00000399744.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22000218.hg.1 |
ENST00000399746.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22000218.hg.1 |
ENST00000428061.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22000218.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22000218.hg.1 |
NM_001199319.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22000218.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22000215.hg.1 |
TC22001451.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22000215.hg.1 |
ENST00000399744.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22000215.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167068.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167068.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167068.hg.1 |
ENST00000428061.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167068.hg.1 |
ENST00000610387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167068.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167068.hg.1 |
NM_001199319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167068.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167068.hg.1 |
TC2200009186.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200167068.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167068.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167068.hg.1 |
ENST00000428061.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167068.hg.1 |
ENST00000610387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167068.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167068.hg.1 |
NM_001199319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167068.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167065.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167065.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167065.hg.1 |
TC2200009186.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200167065.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167065.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000329627.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000399744.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000428061.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000428061.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000428061.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000428061.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000428061.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000450866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000610387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000610387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000610387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000610387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000610387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000610387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000610387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000610387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000610387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
ENST00000610387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_001127649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_001199319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_001199319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_001199319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_001199319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_001199319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_001199319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_001199319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_001199319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_001199319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_001199319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_017929.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009186.hg.1 |
NM_017929.5 |
Refseq |