Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010904.hg.1 |
ENST00000354464.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010904.hg.1 |
ENST00000399536.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010904.hg.1 |
ENST00000561419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010904.hg.1 |
NM_024658.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010904.hg.1 |
TC14002339.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400181695.hg.1 |
ENST00000354464.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181695.hg.1 |
ENST00000558046.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181695.hg.1 |
ENST00000558780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181695.hg.1 |
ENST00000559588.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181695.hg.1 |
ENST00000560155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181695.hg.1 |
ENST00000560798.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181695.hg.1 |
ENST00000561090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181695.hg.1 |
ENST00000561199.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181695.hg.1 |
NM_024658.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400181695.hg.1 |
NR_051979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400181695.hg.1 |
TC1400010722.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400181695.hg.1 |
ENST00000560798.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181695.hg.1 |
ENST00000354464.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181695.hg.1 |
ENST00000558046.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181695.hg.1 |
ENST00000558780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181695.hg.1 |
ENST00000559588.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181695.hg.1 |
ENST00000560155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181695.hg.1 |
ENST00000561090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181695.hg.1 |
ENST00000561199.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181695.hg.1 |
NM_024658.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400181695.hg.1 |
NR_051979.1 |
Refseq |