Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014431.1 | NP_001014431.1 | 2 - 3 | 383 | 67 | |
NM_001014432.1 | NP_001014432.1 | 3 - 4 | 467 | 67 | ||
NM_005163.2 | NP_005154.2 | 2 - 3 | 597 | 67 | ||
XM_005267401.1 | XP_005267458.1 | 2 - 3 | 532 | 67 | ||
XM_017021075.1 | XP_016876564.1 | 3 - 4 | 710 | 67 | ||
XM_017021076.1 | XP_016876565.1 | 3 - 4 | 616 | 67 | ||
XM_017021077.1 | XP_016876566.1 | 2 - 3 | 532 | 67 | ||
XM_017021078.1 | XP_016876567.1 | 3 - 4 | 243 | 67 | ||
GenBank mRNA | AB451242.1 | - | 1 - 2 | 43 | 67 | |
AB451367.1 | - | 1 - 2 | 43 | 67 | ||
AK122894.1 | - | 2 - 3 | 1524 | 67 | ||
AK314256.1 | - | 2 - 3 | 196 | 67 | ||
BC000479.2 | - | 2 - 3 | 318 | 2964603 | 67 | |
BC084538.1 | - | 3 - 4 | 461 | 6645486 | 67 | |
BX648205.1 | - | 1 - 2 | 1001 | 67 | ||
KF836747.1 | - | 3 - 4 | 361 | 67 | ||
KF836748.1 | - | 3 - 4 | 336 | 67 | ||
KF836750.1 | - | 2 - 3 | 183 | 67 | ||
KU177898.1 | - | 1 - 2 | 43 | 67 | ||
KU177899.1 | - | 1 - 2 | 43 | 67 | ||
M63167.1 | - | 2 - 3 | 241 | 67 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |