Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAIAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006340.2 | NP_006331.1 | 13 - 14 | 1643 | 66 | |
XM_005256945.1 | XP_005257002.1 | 14 - 15 | 1742 | 66 | ||
XM_006721636.1 | XP_006721699.1 | 14 - 15 | 1745 | 66 | ||
XM_011524193.1 | XP_011522495.1 | 14 - 15 | 1745 | 66 | ||
GenBank mRNA | AB015020.1 | - | 13 - 14 | 1628 | 66 | |
AK056888.1 | - | 9 - 10 | 1815 | 66 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |