Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00929172_m1
          See other ATG13 GE Assays ›
          Gene Symbol:
          ATG13
          Gene Name:
          autophagy related 13
          Gene Aliases:
          KIAA0652, PARATARG8
          Chromosome Location:
          Chr.11: 46617276 - 46676019 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00929172_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ATG13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001142673.2 NP_001136145.1 3 - 4 750 72
          NM_001205119.1 NP_001192048.1 2 - 3 694 72
          NM_001205120.1 NP_001192049.1 2 - 3 694 72
          NM_001205121.1 NP_001192050.1 3 - 4 745 72
          NM_014741.4 NP_055556.2 3 - 4 750 72
          XM_005253262.2 XP_005253319.1 3 - 4 490 72
          XM_005253263.2 XP_005253320.1 3 - 4 781 72
          XM_005253265.2 XP_005253322.1 4 - 5 987 72
          XM_005253266.2 XP_005253323.1 3 - 4 716 72
          XM_005253268.2 XP_005253325.1 1 - 2 93 72
          XM_006718394.2 XP_006718457.1 4 - 5 837 72
          XM_006718395.2 XP_006718458.1 4 - 5 772 72
          XM_006718396.2 XP_006718459.1 3 - 4 871 72
          XM_011520493.1 XP_011518795.1 5 - 6 974 72
          XM_011520496.2 XP_011518798.1 2 - 3 749 72
          XM_011520499.2 XP_011518801.1 2 - 3 363 72
          XM_011520500.1 XP_011518802.1 4 - 5 837 72
          XM_011520501.1 XP_011518803.1 4 - 5 837 72
          XM_011520502.1 XP_011518804.1 4 - 5 837 72
          XM_011520503.1 XP_011518805.1 4 - 5 837 72
          XM_017018599.1 XP_016874088.1 4 - 5 922 72
          XM_017018600.1 XP_016874089.1 3 - 4 490 72
          XM_017018601.1 XP_016874090.1 2 - 3 665 72
          XM_017018602.1 XP_016874091.1 4 - 5 987 72
          XM_017018603.1 XP_016874092.1 5 - 6 1104 72
          XM_017018604.1 XP_016874093.1 3 - 4 781 72
          XM_017018605.1 XP_016874094.1 4 - 5 772 72
          XM_017018606.1 XP_016874095.1 3 - 4 716 72
          XM_017018607.1 XP_016874096.1 1 - 2 93 72
          XM_017018608.1 XP_016874097.1 3 - 4 490 72
          XM_017018609.1 XP_016874098.1 3 - 4 781 72
          XM_017018610.1 XP_016874099.1 2 - 3 665 72
          XM_017018611.1 XP_016874100.1 3 - 4 716 72
          XM_017018612.1 XP_016874101.1 4 - 5 987 72
          XM_017018613.1 XP_016874102.1 3 - 4 781 72
          XM_017018614.1 XP_016874103.1 4 - 5 772 72
          XM_017018615.1 XP_016874104.1 3 - 4 871 72
          XM_017018616.1 XP_016874105.1 2 - 3 665 72
          XM_017018617.1 XP_016874106.1 4 - 5 870 72
          XM_017018618.1 XP_016874107.1 4 - 5 837 72
          XM_017018619.1 XP_016874108.1 3 - 4 721 72
          XM_017018620.1 XP_016874109.1 3 - 4 781 72
          XM_017018621.1 XP_016874110.1 2 - 3 665 72
          XM_017018622.1 XP_016874111.1 3 - 4 733 72
          GenBank mRNA AB014552.1 - 2 - 3 374 72
          AK023867.1 - 3 - 4 483 72
          AK223603.1 - 2 - 3 370 72
          AK289641.1 - 3 - 4 297 72
          AK293109.1 - 3 - 4 449 72
          BC001331.1 - 3 - 4 179 3458359 72
          BC002378.2 - 3 - 4 449 2961127 72
          BC006191.2 - 3 - 4 491 3936851 72
          CD675643.1 - 1 - 2 69 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100007457.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11000399.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the XM_017018618.1, XM_017018619.1, XM_017018620.1, XM_017018621.1, XM_017018622.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ATG13

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagosome assembly
          macroautophagy
          positive regulation of mitophagy in response to mitochondrial depolarization
          positive regulation of protein targeting to mitochondrion
          protein binding
          protein kinase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR11005013.hg.1 ENST00000312040.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005013.hg.1 ENST00000395549.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005013.hg.1 ENST00000524625.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005013.hg.1 NM_001142673.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11005013.hg.1 NM_014741.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11005013.hg.1 TC11000399.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR11005016.hg.1 ENST00000312040.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005016.hg.1 ENST00000359513.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005016.hg.1 ENST00000395549.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005016.hg.1 ENST00000434074.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005016.hg.1 ENST00000451945.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005016.hg.1 ENST00000524625.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005016.hg.1 ENST00000526508.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005016.hg.1 ENST00000528494.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005016.hg.1 ENST00000529655.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005016.hg.1 ENST00000530500.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005016.hg.1 NM_001142673.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11005016.hg.1 NM_001205119.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11005016.hg.1 NM_001205120.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11005016.hg.1 NM_001205121.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11005016.hg.1 NM_001205122.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11005016.hg.1 NM_014741.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11005016.hg.1 TC11000399.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1100152863.hg.1 ENST00000524625.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100152863.hg.1 ENST00000533325.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100152863.hg.1 ENST00000577256.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100152863.hg.1 ENST00000578626.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100152863.hg.1 ENST00000581416.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100152863.hg.1 NM_001142673.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100152863.hg.1 NM_014741.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100152863.hg.1 TC1100007457.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100152863.hg.1 ENST00000533325.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100152863.hg.1 ENST00000524625.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100152863.hg.1 ENST00000577256.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100152863.hg.1 ENST00000578626.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100152863.hg.1 ENST00000581416.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100152863.hg.1 NM_001142673.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100152863.hg.1 NM_014741.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0