Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00937387_m1
          See other CAT GE Assays ›
          Gene Symbol:
          CAT
          Gene Name:
          catalase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 34438925 - 34472060 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00937387_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CAT
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001752.3 NP_001743.1 10 - 11 1415 72
          GenBank mRNA AK291585.1 - 10 - 11 1415 72
          AK301577.1 - 9 - 10 1185 72
          AY028632.1 - 10 - 11 1326 72
          BC027300.1 - 3 - 4 429 72
          BC110398.1 - 10 - 11 1404 5551309 72
          BC112217.1 - 10 - 11 1364 8327685 72
          BC112219.1 - 10 - 11 1364 8327687 72
          X04076.1 - 10 - 11 1396 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100007257.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11000327.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CAT

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          osteoblast differentiation
          ureteric bud development
          response to hypoxia
          purine nucleotide catabolic process
          triglyceride metabolic process
          aging
          cholesterol metabolic process
          aerobic respiration
          response to xenobiotic stimulus
          response to light intensity
          UV protection
          response to ozone
          response to lead ion
          positive regulation of phosphatidylinositol 3-kinase signaling
          response to activity
          response to inactivity
          hemoglobin metabolic process
          negative regulation of NF-kappaB transcription factor activity
          response to estradiol
          response to insulin
          response to vitamin A
          response to vitamin E
          response to L-ascorbic acid
          response to hydrogen peroxide
          hydrogen peroxide catabolic process
          negative regulation of apoptotic process
          response to ethanol
          response to cadmium ion
          positive regulation of NF-kappaB transcription factor activity
          protein tetramerization
          protein homotetramerization
          positive regulation of cell division
          response to hyperoxia
          response to fatty acid
          cellular response to growth factor stimulus
          response to phenylpropanoid
          cellular oxidant detoxification
          aminoacylase activity
          catalase activity
          signaling receptor binding
          antioxidant activity
          oxidoreductase activity, acting on peroxide as acceptor
          enzyme binding
          heme binding
          protein homodimerization activity
          metal ion binding
          NADP binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR11004196.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11004196.hg.1 NM_001752.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11004196.hg.1 TC11000327.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000525707.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000525707.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000525707.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000525707.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000525707.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000525707.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000528104.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000528104.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000528104.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000530343.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000530343.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000530343.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000530343.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000534710.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000534710.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000534710.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000534710.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000534710.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 ENST00000534710.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100151165.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100151165.hg.1 ENST00000525707.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100151165.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100151165.hg.1 TC1100007257.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100151165.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100151165.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100151165.hg.1 ENST00000525707.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100151164.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100151164.hg.1 ENST00000525707.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100151164.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100151164.hg.1 TC1100007257.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100151164.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100151164.hg.1 ENST00000525707.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100151164.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000241052.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000525707.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000525707.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000525707.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000525707.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000525707.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000525707.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000528104.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000528104.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000528104.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000530343.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000530343.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000530343.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000530343.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000534710.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000534710.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000534710.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000534710.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000534710.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 ENST00000534710.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 NM_001752.3 Refseq
          Affymetrix Human Clariom D Assay TC1100007257.hg.1 NM_001752.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix