Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012786.hg.1 |
TC20000928.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012786.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012786.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000165718.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165718.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000165718.hg.1 |
TC2000009392.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000165718.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000165718.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |