Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00941958_m1
          See other SYNE1 GE Assays ›
          Gene Symbol:
          SYNE1
          Gene Name:
          spectrin repeat containing nuclear envelope protein 1
          Gene Aliases:
          8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
          Chromosome Location:
          Chr.6: 152121684 - 152637399 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00941958_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SYNE1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033071.3 NP_149062.1 132 - 133 24350 106
          NM_182961.3 NP_892006.3 133 - 134 24728 106
          XM_006715407.1 XP_006715470.1 133 - 134 24641 106
          XM_006715408.2 XP_006715471.1 133 - 134 24633 106
          XM_006715409.1 XP_006715472.1 132 - 133 24620 106
          XM_006715410.2 XP_006715473.1 133 - 134 24645 106
          XM_006715411.1 XP_006715474.1 132 - 133 24590 106
          XM_006715412.2 XP_006715475.1 133 - 134 24630 106
          XM_006715413.2 XP_006715476.1 133 - 134 24645 106
          XM_006715414.1 XP_006715477.1 131 - 132 24569 106
          XM_006715415.2 XP_006715478.1 133 - 134 24645 106
          XM_006715416.2 XP_006715479.1 133 - 134 24630 106
          XM_006715417.2 XP_006715480.1 132 - 133 24504 106
          XM_006715420.2 XP_006715483.1 132 - 133 24492 106
          XM_006715421.2 XP_006715484.1 132 - 133 24489 106
          XM_006715422.1 XP_006715485.1 132 - 133 24482 106
          XM_006715423.2 XP_006715486.1 133 - 134 24645 106
          XM_006715424.2 XP_006715487.1 133 - 134 24645 106
          XM_006715425.2 XP_006715488.1 133 - 134 24645 106
          XM_011535641.2 XP_011533943.1 133 - 134 24642 106
          XM_011535642.2 XP_011533944.1 133 - 134 24630 106
          XM_011535643.1 XP_011533945.1 132 - 133 24476 106
          XM_011535644.1 XP_011533946.1 119 - 120 22692 106
          XM_011535645.2 XP_011533947.1 115 - 116 22085 106
          XM_017010608.1 XP_016866097.1 134 - 135 24760 106
          XM_017010609.1 XP_016866098.1 133 - 134 24567 106
          XM_017010610.1 XP_016866099.1 132 - 133 24546 106
          XM_017010611.1 XP_016866100.1 132 - 133 24349 106
          XM_017010612.1 XP_016866101.1 131 - 132 24273 106
          XM_017010613.1 XP_016866102.1 133 - 134 24642 106
          XM_017010614.1 XP_016866103.1 132 - 133 24489 106
          XM_017010615.1 XP_016866104.1 132 - 133 24489 106
          XM_017010616.1 XP_016866105.1 133 - 134 24645 106
          XM_017010617.1 XP_016866106.1 133 - 134 24642 106
          XM_017010618.1 XP_016866107.1 133 - 134 24630 106
          XM_017010619.1 XP_016866108.1 119 - 120 22724 106
          GenBank mRNA AB018339.2 - 21 - 22 3365 106
          AF444779.1 - 8 - 9 1238 106
          AF495910.1 - 133 - 134 24728 106
          AF535142.1 - 132 - 133 24350 106
          AK122691.1 - 13 - 14 2008 106
          AK127076.1 - 17 - 18 2705 106
          AK304825.1 - 7 - 8 930 106
          AK316101.1 - 6 - 7 945 106
          AY061755.1 - 48 - 49 7818 106
          AY061756.1 - 5 - 6 808 106
          AY184203.1 - 4 - 5 673 106
          AY184206.1 - 46 - 47 7713 106
          BC090927.1 - 6 - 7 990 106
          BX537837.1 - 15 - 16 2306 106
          BX647837.1 - 18 - 19 2900 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600013588.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06002238.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SYNE1

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleus organization
          Golgi organization
          nuclear migration
          muscle cell differentiation
          nuclear matrix anchoring at nuclear membrane
          RNA binding
          actin binding
          protein binding
          lamin binding
          protein homodimerization activity
          actin filament binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR06027934.hg.1 ENST00000265368.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06027934.hg.1 ENST00000341594.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06027934.hg.1 ENST00000347037.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06027934.hg.1 ENST00000354674.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06027934.hg.1 ENST00000356820.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06027934.hg.1 ENST00000367247.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06027934.hg.1 ENST00000367255.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06027934.hg.1 ENST00000423061.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06027934.hg.1 ENST00000448038.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06027934.hg.1 ENST00000539504.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06027934.hg.1 NM_033071.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06027934.hg.1 NM_182961.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06027934.hg.1 TC06002238.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06027933.hg.1 ENST00000265368.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06027933.hg.1 ENST00000341594.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06027933.hg.1 ENST00000347037.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06027933.hg.1 ENST00000354674.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06027933.hg.1 ENST00000356820.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06027933.hg.1 ENST00000367247.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06027933.hg.1 ENST00000367255.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06027933.hg.1 ENST00000423061.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06027933.hg.1 ENST00000448038.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06027933.hg.1 ENST00000539504.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06027933.hg.1 NM_033071.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06027933.hg.1 NM_182961.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06027933.hg.1 TC06002238.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000341594.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000347037.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000354674.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000367251.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000367255.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000367256.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000367257.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000409694.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000423061.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000460912.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000476519.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000490866.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000536990.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000539504.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 NM_033071.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 NM_182961.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 TC0600013588.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000354674.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000341594.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000347037.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000367251.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000367255.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000367256.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000367257.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000409694.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000423061.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000460912.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000476519.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000490866.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000536990.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 ENST00000539504.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 NM_033071.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600193779.hg.1 NM_182961.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000341594.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000347037.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000354674.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000367251.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000367255.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000367256.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000367257.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000409694.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000423061.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000460912.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000476519.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000536990.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000539504.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 NM_033071.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 NM_182961.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 TC0600013588.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000341594.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000347037.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000354674.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000367251.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000367255.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000367256.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000367257.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000409694.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000423061.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000460912.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000476519.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000536990.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 ENST00000539504.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 NM_033071.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600193778.hg.1 NM_182961.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0