Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00942013_m1
          See other SYNE1 GE Assays ›
          Gene Symbol:
          SYNE1
          Gene Name:
          spectrin repeat containing nuclear envelope protein 1
          Gene Aliases:
          8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
          Chromosome Location:
          Chr.6: 152121684 - 152637399 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00942013_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SYNE1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033071.3 NP_149062.1 51 - 52 8169 110
          NM_182961.3 NP_892006.3 51 - 52 8313 110
          XM_006715407.1 XP_006715470.1 51 - 52 8211 110
          XM_006715408.2 XP_006715471.1 51 - 52 8215 110
          XM_006715409.1 XP_006715472.1 50 - 51 8190 110
          XM_006715410.2 XP_006715473.1 51 - 52 8215 110
          XM_006715411.1 XP_006715474.1 50 - 51 8160 110
          XM_006715412.2 XP_006715475.1 51 - 52 8215 110
          XM_006715413.2 XP_006715476.1 51 - 52 8215 110
          XM_006715414.1 XP_006715477.1 49 - 50 8139 110
          XM_006715415.2 XP_006715478.1 51 - 52 8215 110
          XM_006715416.2 XP_006715479.1 51 - 52 8215 110
          XM_006715417.2 XP_006715480.1 51 - 52 8215 110
          XM_006715420.2 XP_006715483.1 51 - 52 8215 110
          XM_006715421.2 XP_006715484.1 51 - 52 8215 110
          XM_006715422.1 XP_006715485.1 50 - 51 8052 110
          XM_006715423.2 XP_006715486.1 51 - 52 8215 110
          XM_006715424.2 XP_006715487.1 51 - 52 8215 110
          XM_006715425.2 XP_006715488.1 51 - 52 8215 110
          XM_011535641.2 XP_011533943.1 51 - 52 8215 110
          XM_011535642.2 XP_011533944.1 51 - 52 8215 110
          XM_011535643.1 XP_011533945.1 50 - 51 8046 110
          XM_011535644.1 XP_011533946.1 37 - 38 6262 110
          XM_011535645.2 XP_011533947.1 33 - 34 5655 110
          XM_017010608.1 XP_016866097.1 52 - 53 8330 110
          XM_017010609.1 XP_016866098.1 51 - 52 8137 110
          XM_017010610.1 XP_016866099.1 50 - 51 8116 110
          XM_017010611.1 XP_016866100.1 50 - 51 7919 110
          XM_017010612.1 XP_016866101.1 49 - 50 7843 110
          XM_017010613.1 XP_016866102.1 51 - 52 8215 110
          XM_017010614.1 XP_016866103.1 51 - 52 8215 110
          XM_017010615.1 XP_016866104.1 51 - 52 8215 110
          XM_017010616.1 XP_016866105.1 51 - 52 8215 110
          XM_017010617.1 XP_016866106.1 51 - 52 8215 110
          XM_017010618.1 XP_016866107.1 51 - 52 8215 110
          XM_017010619.1 XP_016866108.1 37 - 38 6294 110
          GenBank mRNA AF495910.1 - 51 - 52 8313 110
          AF535142.1 - 51 - 52 8169 110
          FM162565.1 - 49 - 50 7929 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600013588.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06002238.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SYNE1

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleus organization
          Golgi organization
          nuclear migration
          muscle cell differentiation
          nuclear matrix anchoring at nuclear membrane
          RNA binding
          actin binding
          protein binding
          lamin binding
          protein homodimerization activity
          actin filament binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR06028058.hg.1 ENST00000265368.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06028058.hg.1 ENST00000341594.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06028058.hg.1 ENST00000367255.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06028058.hg.1 ENST00000423061.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06028058.hg.1 ENST00000448038.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06028058.hg.1 NM_033071.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06028058.hg.1 NM_182961.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06028058.hg.1 TC06002238.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0600193908.hg.1 ENST00000341594.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193908.hg.1 ENST00000367255.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193908.hg.1 ENST00000423061.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193908.hg.1 ENST00000461872.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193908.hg.1 NM_033071.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600193908.hg.1 NM_182961.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600193908.hg.1 TC0600013588.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600193908.hg.1 ENST00000341594.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193908.hg.1 ENST00000367255.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193908.hg.1 ENST00000423061.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193908.hg.1 ENST00000461872.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600193908.hg.1 NM_033071.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600193908.hg.1 NM_182961.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0