Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00942070_m1
          See other SYNE1 GE Assays ›
          Gene Symbol:
          SYNE1
          Gene Name:
          spectrin repeat containing nuclear envelope protein 1
          Gene Aliases:
          8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
          Chromosome Location:
          Chr.6: 152121684 - 152637399 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00942070_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SYNE1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006715407.1 XP_006715470.1 143 - 144 26282 67
          XM_006715408.2 XP_006715471.1 143 - 144 26274 67
          XM_006715409.1 XP_006715472.1 142 - 143 26261 67
          XM_006715411.1 XP_006715474.1 142 - 143 26231 67
          XM_006715413.2 XP_006715476.1 142 - 143 26217 67
          XM_006715414.1 XP_006715477.1 141 - 142 26210 67
          XM_006715417.2 XP_006715480.1 142 - 143 26145 67
          XM_006715420.2 XP_006715483.1 142 - 143 26133 67
          XM_006715421.2 XP_006715484.1 142 - 143 26130 67
          XM_006715422.1 XP_006715485.1 142 - 143 26123 67
          XM_006715423.2 XP_006715486.1 143 - 144 26286 67
          XM_011535641.2 XP_011533943.1 143 - 144 26283 67
          XM_011535642.2 XP_011533944.1 143 - 144 26271 67
          XM_011535643.1 XP_011533945.1 142 - 143 26117 67
          XM_011535644.1 XP_011533946.1 129 - 130 24333 67
          XM_011535645.2 XP_011533947.1 125 - 126 23726 67
          XM_017010608.1 XP_016866097.1 144 - 145 26401 67
          XM_017010609.1 XP_016866098.1 143 - 144 26208 67
          XM_017010610.1 XP_016866099.1 142 - 143 26187 67
          XM_017010611.1 XP_016866100.1 142 - 143 25990 67
          XM_017010612.1 XP_016866101.1 141 - 142 25914 67
          XM_017010614.1 XP_016866103.1 142 - 143 26130 67
          XM_017010616.1 XP_016866105.1 142 - 143 26217 67
          XM_017010619.1 XP_016866108.1 129 - 130 24365 67

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleus organization
          Golgi organization
          nuclear migration
          muscle cell differentiation
          nuclear matrix anchoring at nuclear membrane
          RNA binding
          actin binding
          protein binding
          lamin binding
          protein homodimerization activity
          actin filament binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0