Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00945963_s1
          See other LINC00260 GE Assays ›
          Gene Symbol:
          LINC00260
          Gene Name:
          long intergenic non-protein coding RNA 260
          Gene Aliases:
          C1orf217, NCRNA00260
          Chromosome Location:
          Chr.1: 203730577 - 203731853 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00945963_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00260
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046213.1 1 835 92
          GenBank mRNA BC000988.2 - 1 835 3450898 92
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DB319348.1 - 1 Chr. 1 - 203731312 - 203731849 [-] on Build GRCh38 92

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream small nucleolar RNA,
          H/ACA box 77
          SNORA77 ACA63,
          SNORA77A
          -872.0 Sense Chr. 1 - 203729581 - 203729705 on NCBI Build GRCh38
          Downstream lymphocyte transmembrane adaptor 1 LAX1 LAX 33.3kb Sense Chr. 1 - 203765132 - 203778175 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3210810 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P23234 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 12848 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 27512 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3210810 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P23234 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P23234 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array XR_040170_at - -
          Affymetrix Human Gene 1.0 ST Array XR_040168_at - -
          Affymetrix Human Gene 1.0 ST Array XR_040169_at - -
          Affymetrix Human Gene 1.0 ST Array 7908967 - -
          Affymetrix Human Gene 1.1 ST Array 7908966 - -
          Affymetrix HT HG-U133+ PM Array Plate 210711_PM_at - -
          Affymetrix Human Genome U219 Array 11760566_at - -
          Affymetrix Human Genome U219 Array 11760566_PM_at - -
          Human Genome U133 Plus 2.0 Array 210711_at - -
          Human Genome U133 Plus 2.0 Array 210711_PM_at - -
          Human Genome U133A 2.0 Array 210711_at - -
          Human Genome U133A 2.0 Array 210711_PM_at - -
          AffymetrixPathwork Diagnostics Pathchip pw541257 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210711_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210711_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210711_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210711_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210711_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210711_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210711_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210711_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210711_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210711_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210711_at7 - -
          Human Genome U133 Set 210711_at - -
          Human Genome U133 Set 210711_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 105813 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1733412 BC000988.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1733412 BC000988.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1733412 BC000988.2 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0