Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00952266_m1
          See other ARHGAP18 GE Assays ›
          Gene Symbol:
          ARHGAP18
          Gene Name:
          Rho GTPase activating protein 18
          Gene Aliases:
          MacGAP, SENEX, bA307O14.2
          Chromosome Location:
          Chr.6: 129576145 - 129710225 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00952266_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ARHGAP18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033515.2 NP_277050.2 10 - 11 1454 113
          XM_005267212.2 XP_005267269.2 10 - 11 1530 113
          XM_005267213.1 XP_005267270.1 10 - 11 1454 113
          XM_017011494.1 XP_016866983.1 10 - 11 1530 113
          GenBank mRNA AB053293.1 - 9 - 10 1252 113
          AL834511.1 - 10 - 11 1392 113
          AY643712.1 - 10 - 11 1455 113
          BC101708.1 - 10 - 11 1448 8069214 113
          BC107416.1 - 10 - 11 1410 4040931 113
          BC111940.1 - 10 - 11 1448 8327408 113
          BC143531.1 - 10 - 11 1448 113

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600013147.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06004137.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ARHGAP18

          Gene Ontology Categories:

          Function(s) Process(es)

          small GTPase mediated signal transduction
          regulation of cell shape
          regulation of actin filament polymerization
          regulation of actin cytoskeleton organization
          positive regulation of GTPase activity
          regulation of small GTPase mediated signal transduction
          cell-cell adhesion
          regulation of cell motility
          GTPase activator activity
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000463225.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000463225.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000463225.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000463225.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000463225.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000463225.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000483367.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000483367.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000483367.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000483367.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000483367.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 ENST00000483367.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06026301.hg.1 ENST00000275189.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06026301.hg.1 ENST00000368149.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06026301.hg.1 NM_033515.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06026301.hg.1 TC06004137.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0600190711.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600190711.hg.1 NM_033515.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600190711.hg.1 TC0600013147.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600190711.hg.1 NM_033515.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600190711.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000368149.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000463225.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000463225.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000463225.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000463225.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000463225.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000463225.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000483367.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000483367.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000483367.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000483367.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000483367.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 ENST00000483367.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 NM_033515.2 Refseq
          Affymetrix Human Clariom D Assay TC0600013147.hg.1 NM_033515.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0