Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206927.1 | NP_001193856.1 | 82 - 83 | 12551 | 80 | |
XM_011514318.2 | XP_011512620.1 | 81 - 82 | 12499 | 80 | ||
XM_011514319.2 | XP_011512621.1 | 81 - 82 | 12454 | 80 | ||
XM_011514320.2 | XP_011512622.1 | 80 - 81 | 12325 | 80 | ||
XM_011514321.1 | XP_011512623.1 | 79 - 80 | 11909 | 80 | ||
XM_017010325.1 | XP_016865814.1 | 82 - 83 | 12562 | 80 | ||
GenBank mRNA | AF356519.1 | - | 80 - 81 | 12045 | 80 | |
AK093434.1 | - | 2 - 3 | 207 | 80 | ||
AK093653.1 | - | 8 - 9 | 1273 | 80 |
Set Membership: |
![]() ![]() ![]() |