Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00954390_m1
          See other ADRA1A GE Assays ›
          Gene Symbol:
          ADRA1A
          Gene Name:
          adrenoceptor alpha 1A
          Gene Aliases:
          ADRA1C, ADRA1L1, ALPHA1AAR
          Chromosome Location:
          Chr.8: 26738113 - 26870994 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00954390_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ADRA1A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000680.3 NP_000671.2 1 - 2 312 159
          XM_006716292.3 XP_006716355.1 1 - 2 339 159
          XM_006716293.3 XP_006716356.1 1 - 2 339 159
          XM_011544411.2 XP_011542713.1 1 - 2 339 159
          XM_011544412.2 XP_011542714.1 1 - 2 339 159
          XM_017013094.1 XP_016868583.1 2 - 3 3899 159
          XM_017013095.1 XP_016868584.1 2 - 3 3922 159
          XM_017013096.1 XP_016868585.1 1 - 2 339 159
          GenBank mRNA AK289548.1 - 1 - 2 313 159
          AK311558.1 - 2 283 159

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800012393.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08001076.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK289548.1, NM_000680.3, XM_006716292.3, XM_006716293.3, XM_011544411.2, XM_011544412.2, XM_017013094.1, XM_017013095.1, XM_017013096.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ADRA1A

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of heart rate involved in baroreceptor response to increased systemic arterial blood pressure
          norepinephrine-epinephrine vasoconstriction involved in regulation of systemic arterial blood pressure
          positive regulation of heart rate by epinephrine-norepinephrine
          positive regulation of the force of heart contraction by epinephrine-norepinephrine
          positive regulation of systemic arterial blood pressure
          apoptotic process
          smooth muscle contraction
          signal transduction
          G protein-coupled receptor signaling pathway
          phospholipase C-activating G protein-coupled receptor signaling pathway
          activation of phospholipase C activity
          positive regulation of cytosolic calcium ion concentration
          cell-cell signaling
          adult heart development
          aging
          negative regulation of cell population proliferation
          response to xenobiotic stimulus
          response to hormone
          cell growth
          positive regulation of synaptic transmission, GABAergic
          negative regulation of Rho protein signal transduction
          organ growth
          intracellular signal transduction
          positive regulation of MAPK cascade
          positive regulation of action potential
          positive regulation of vasoconstriction
          positive regulation of smooth muscle contraction
          micturition
          calcium ion transport into cytosol
          positive regulation of cardiac muscle contraction
          positive regulation of ERK1 and ERK2 cascade
          adenylate cyclase-activating adrenergic receptor signaling pathway
          positive regulation of protein kinase C signaling
          pilomotor reflex
          alpha1-adrenergic receptor activity
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR08013719.hg.1 ENST00000276393.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013719.hg.1 ENST00000354550.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013719.hg.1 ENST00000358857.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013719.hg.1 ENST00000380572.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013719.hg.1 ENST00000380573.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013719.hg.1 ENST00000380581.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013719.hg.1 ENST00000380582.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013719.hg.1 ENST00000380586.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013719.hg.1 ENST00000380587.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013719.hg.1 ENST00000519229.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013719.hg.1 NM_000680.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08013719.hg.1 NM_033302.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08013719.hg.1 NM_033303.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08013719.hg.1 NM_033304.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08013719.hg.1 TC08001076.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08013720.hg.1 ENST00000276393.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013720.hg.1 ENST00000354550.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013720.hg.1 ENST00000358857.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013720.hg.1 ENST00000380572.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013720.hg.1 ENST00000380573.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013720.hg.1 ENST00000380581.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013720.hg.1 ENST00000380582.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013720.hg.1 ENST00000380586.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013720.hg.1 ENST00000380587.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013720.hg.1 NM_000680.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08013720.hg.1 NM_033302.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08013720.hg.1 NM_033303.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08013720.hg.1 NM_033304.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08013720.hg.1 TC08001076.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08013721.hg.1 ENST00000276393.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013721.hg.1 ENST00000358857.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013721.hg.1 ENST00000380572.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013721.hg.1 ENST00000380573.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013721.hg.1 ENST00000380581.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013721.hg.1 ENST00000380582.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013721.hg.1 ENST00000380586.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013721.hg.1 ENST00000380587.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013721.hg.1 NM_000680.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08013721.hg.1 NM_033302.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08013721.hg.1 NM_033303.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08013721.hg.1 NM_033304.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08013721.hg.1 TC08001076.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0800186820.hg.1 ENST00000276393.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186820.hg.1 ENST00000380572.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186820.hg.1 ENST00000380582.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186820.hg.1 ENST00000380586.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186820.hg.1 ENST00000519096.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186820.hg.1 ENST00000521711.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186820.hg.1 NM_000680.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800186820.hg.1 NM_033302.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800186820.hg.1 NM_033303.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800186820.hg.1 NM_033304.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800186820.hg.1 TC0800012393.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800186820.hg.1 ENST00000276393.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186820.hg.1 ENST00000380572.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186820.hg.1 ENST00000380582.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186820.hg.1 ENST00000380586.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186820.hg.1 ENST00000519096.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186820.hg.1 ENST00000521711.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186820.hg.1 NM_000680.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800186820.hg.1 NM_033302.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800186820.hg.1 NM_033303.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800186820.hg.1 NM_033304.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 ENST00000276393.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 ENST00000354550.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 ENST00000380572.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 ENST00000380582.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 ENST00000380586.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 ENST00000519096.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 ENST00000519229.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 ENST00000521711.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 NM_000680.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 NM_033302.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 NM_033303.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 NM_033304.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 TC0800012393.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 ENST00000276393.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 ENST00000354550.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 ENST00000380572.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 ENST00000380582.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 ENST00000380586.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 ENST00000519096.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 ENST00000519229.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 ENST00000521711.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 NM_000680.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 NM_033302.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 NM_033303.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800186817.hg.1 NM_033304.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800186819.hg.1 ENST00000276393.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186819.hg.1 ENST00000354550.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186819.hg.1 ENST00000380572.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186819.hg.1 ENST00000380582.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186819.hg.1 ENST00000380586.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186819.hg.1 ENST00000519096.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186819.hg.1 ENST00000521711.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186819.hg.1 NM_000680.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800186819.hg.1 NM_033302.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800186819.hg.1 NM_033303.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800186819.hg.1 NM_033304.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800186819.hg.1 TC0800012393.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800186819.hg.1 ENST00000276393.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186819.hg.1 ENST00000354550.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186819.hg.1 ENST00000380572.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186819.hg.1 ENST00000380582.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186819.hg.1 ENST00000380586.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186819.hg.1 ENST00000519096.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186819.hg.1 ENST00000521711.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800186819.hg.1 NM_000680.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800186819.hg.1 NM_033302.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800186819.hg.1 NM_033303.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800186819.hg.1 NM_033304.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix