Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00961200_m1
          See other TNKS2 GE Assays ›
          Gene Symbol:
          TNKS2
          Gene Name:
          tankyrase 2
          Gene Aliases:
          ARTD6, PARP-5b, PARP-5c, PARP5B, PARP5C, TANK2, TNKL, pART6
          Chromosome Location:
          Chr.10: 91798394 - 91865475 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00961200_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TNKS2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025235.3 NP_079511.1 3 - 4 813 87
          XM_005270185.4 XP_005270242.2 3 - 4 844 87
          XM_011540213.1 XP_011538515.1 3 - 4 844 87
          XM_017016696.1 XP_016872185.1 3 - 4 781 87
          XM_017016697.1 XP_016872186.1 3 - 4 797 87
          XM_017016698.1 XP_016872187.1 3 - 4 508 87
          XM_017016699.1 XP_016872188.1 3 - 4 797 87
          XM_017016700.1 XP_016872189.1 3 - 4 394 87
          XM_017016701.1 XP_016872190.1 3 - 4 844 87
          GenBank mRNA AF264912.1 - 3 - 4 516 87
          AF305081.1 - 3 - 4 818 87
          AF309033.1 - 3 - 4 767 87
          AF329696.1 - 3 - 4 655 87
          AF342982.1 - 3 - 4 516 87
          AF438201.1 - 3 - 4 895 87
          AK314612.1 - 3 - 4 781 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000008447.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000654.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the XM_017016700.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TNKS2

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          protein ADP-ribosylation
          Wnt signaling pathway
          positive regulation of telomere maintenance via telomerase
          multicellular organism growth
          regulation of multicellular organism growth
          protein localization to chromosome, telomeric region
          protein auto-ADP-ribosylation
          positive regulation of canonical Wnt signaling pathway
          positive regulation of telomere capping
          negative regulation of telomere maintenance via telomere lengthening
          NAD+ ADP-ribosyltransferase activity
          protein binding
          enzyme binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10007266.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10007266.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10007266.hg.1 TC10000654.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10007267.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10007267.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10007267.hg.1 TC10000654.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1000157348.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000157348.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000157348.hg.1 TC1000008447.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000157348.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000157348.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000157350.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000157350.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000157350.hg.1 TC1000008447.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000157350.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000157350.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5df8fb85f4-n94h7:80/100.66.130.62:80.
          git-commit: 8f66f361c5203ebe605e386d8a840ec15b9b8304
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0