Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007272.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007272.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007272.hg.1 |
TC10000654.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157360.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157360.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157360.hg.1 |
TC1000008447.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000157360.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157360.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
ENST00000371627.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008447.hg.1 |
NM_025235.3 |
Refseq |