Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00964974_g1
          See other PSMB3 GE Assays ›
          Gene Symbol:
          PSMB3
          Gene Name:
          proteasome subunit beta 3
          Gene Aliases:
          HC10-II
          Chromosome Location:
          Chr.17: 38752713 - 38764231 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00964974_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PSMB3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002795.3 NP_002786.2 1 - 2 111 72
          NR_104194.1 1 - 2 111 72
          NR_104195.1 1 - 2 111 72
          GenBank mRNA BC013008.2 - 1 - 2 74 3010175 72
          BG942618.1 - 1 - 2 112 72
          BQ428285.1 - 1 - 2 71 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700007720.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17000466.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the BC013008.2, NM_002795.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PSMB3

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          protein polyubiquitination
          stimulatory C-type lectin receptor signaling pathway
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
          regulation of cellular amino acid metabolic process
          viral process
          anaphase-promoting complex-dependent catabolic process
          tumor necrosis factor-mediated signaling pathway
          NIK/NF-kappaB signaling
          Fc-epsilon receptor signaling pathway
          proteasome-mediated ubiquitin-dependent protein catabolic process
          regulation of mRNA stability
          T cell receptor signaling pathway
          Wnt signaling pathway, planar cell polarity pathway
          negative regulation of canonical Wnt signaling pathway
          positive regulation of canonical Wnt signaling pathway
          threonine-type endopeptidase activity
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR17005475.hg.1 ENST00000225426.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17005475.hg.1 NM_002795.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17005475.hg.1 TC17000466.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0200015677.hg.1 ENST00000457237.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17005476.hg.1 ENST00000225426.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17005476.hg.1 NM_002795.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17005476.hg.1 TC17000466.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1200010863.hg.1 ENST00000554248.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17005474.hg.1 ENST00000225426.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17005474.hg.1 NM_002795.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17005474.hg.1 TC17000466.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000610434.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000610434.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000610434.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000610434.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000610434.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000613870.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000613870.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000613870.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000613870.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000613870.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000613870.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000613870.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000613870.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000614132.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000614132.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000614132.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000614132.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000614132.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000614132.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000614132.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000618159.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000618159.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000618159.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000618159.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000619426.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000619426.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000619426.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000619426.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000619426.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000619426.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000619426.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000619426.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000619426.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000620309.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000620309.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000620309.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000620309.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000620309.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 ENST00000620309.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 NM_002795.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 NM_002795.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 NM_002795.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 NM_002795.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 NM_002795.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 NM_002795.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 NM_002795.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 NM_002795.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 NM_002795.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 NR_104194.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 NR_104194.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 NR_104194.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 NR_104194.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 NR_104194.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 NR_104194.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 NR_104195.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 NR_104195.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 NR_104195.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 NR_104195.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 NR_104195.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 NR_104195.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 NR_104195.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007720.hg.1 NR_104195.1 Refseq
          Affymetrix Human Clariom D Assay TC0200015677.hg.1 ENST00000457237.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000610434.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000610434.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000610434.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000610434.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000610434.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000613870.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000613870.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000613870.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000613870.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000613870.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000613870.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000613870.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000613870.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000614132.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000614132.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000614132.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000614132.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000614132.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000614132.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000614132.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000618159.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000618159.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000618159.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000618159.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000619426.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000619426.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000619426.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000619426.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000619426.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000619426.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000619426.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000619426.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000619426.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000620309.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000620309.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000620309.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000620309.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000620309.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 ENST00000620309.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 NM_002795.3 Refseq
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 NM_002795.3 Refseq
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 NM_002795.3 Refseq
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 NM_002795.3 Refseq
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 NM_002795.3 Refseq
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 NM_002795.3 Refseq
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 NM_002795.3 Refseq
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 NM_002795.3 Refseq
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 NM_002795.3 Refseq
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 NR_104194.1 Refseq
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 NR_104194.1 Refseq
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 NR_104194.1 Refseq
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 NR_104194.1 Refseq
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 NR_104194.1 Refseq
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 NR_104194.1 Refseq
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 NR_104195.1 Refseq
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 NR_104195.1 Refseq
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 NR_104195.1 Refseq
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 NR_104195.1 Refseq
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 NR_104195.1 Refseq
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 NR_104195.1 Refseq
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 NR_104195.1 Refseq
          Affymetrix Human Clariom D Assay TC1700007720.hg.1 NR_104195.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010863.hg.1 ENST00000554248.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700155697.hg.1 ENST00000610434.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700155697.hg.1 ENST00000613870.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700155697.hg.1 ENST00000619426.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700155697.hg.1 ENST00000620309.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700155697.hg.1 NM_002795.3 Refseq
          Affymetrix Human Clariom D Assay PSR1700155697.hg.1 NR_104194.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700155697.hg.1 NR_104195.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700155697.hg.1 TC1700007720.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700155697.hg.1 ENST00000613870.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700155697.hg.1 ENST00000610434.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700155697.hg.1 ENST00000619426.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700155697.hg.1 ENST00000620309.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700155697.hg.1 NM_002795.3 Refseq
          Affymetrix Human Clariom D Assay PSR1700155697.hg.1 NR_104194.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700155697.hg.1 NR_104195.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700155700.hg.1 ENST00000610434.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700155700.hg.1 ENST00000613870.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700155700.hg.1 ENST00000619426.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700155700.hg.1 ENST00000620309.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700155700.hg.1 NM_002795.3 Refseq
          Affymetrix Human Clariom D Assay PSR1700155700.hg.1 NR_104194.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700155700.hg.1 NR_104195.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700155700.hg.1 TC1700007720.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700155700.hg.1 ENST00000610434.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700155700.hg.1 ENST00000613870.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700155700.hg.1 ENST00000619426.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700155700.hg.1 ENST00000620309.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700155700.hg.1 NM_002795.3 Refseq
          Affymetrix Human Clariom D Assay PSR1700155700.hg.1 NR_104194.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700155700.hg.1 NR_104195.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix