Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00968801_m1
          See other CAMK2G GE Assays ›
          Gene Symbol:
          CAMK2G
          Gene Name:
          calcium/calmodulin dependent protein kinase II gamma
          Gene Aliases:
          CAMK, CAMK-II, CAMKG
          Chromosome Location:
          Chr.10: 73812501 - 73874591 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00968801_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CAMK2G
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001204492.1 NP_001191421.1 16 - 17 1299 84
          NM_001320898.1 NP_001307827.1 16 - 17 1263 84
          NM_172169.2 NP_751909.1 16 - 17 1263 84
          NM_172170.4 NP_751910.1 15 - 16 1236 84
          NM_172171.2 NP_751911.1 15 - 16 1236 84
          XM_005270195.1 XP_005270252.1 16 - 17 1290 84
          XM_005270197.1 XP_005270254.1 16 - 17 1254 84
          XM_005270200.1 XP_005270257.1 15 - 16 1191 84
          XM_005270203.1 XP_005270260.1 16 - 17 1260 84
          XM_006717993.1 XP_006718056.1 17 - 18 1323 84
          XM_006717996.1 XP_006718059.1 16 - 17 1260 84
          XM_006717997.1 XP_006718060.1 17 - 18 1323 84
          XM_011540225.2 XP_011538527.1 15 - 16 1175 84
          XM_017016716.1 XP_016872205.1 17 - 18 1323 84
          XM_017016717.1 XP_016872206.1 17 - 18 1271 84
          XM_017016718.1 XP_016872207.1 16 - 17 1290 84
          XM_017016720.1 XP_016872209.1 16 - 17 1260 84
          XM_017016721.1 XP_016872210.1 17 - 18 1398 84
          XM_017016723.1 XP_016872212.1 15 - 16 1227 84
          XM_017016725.1 XP_016872214.1 15 - 16 1191 84
          XM_017016728.1 XP_016872217.1 15 - 16 1175 84
          XM_017016729.1 XP_016872218.1 15 - 16 1058 84
          XM_017016731.1 XP_016872220.1 15 - 16 1166 84
          XM_017016732.1 XP_016872221.1 15 - 16 1096 84
          XM_017016733.1 XP_016872222.1 15 - 16 1175 84
          XM_017016734.1 XP_016872223.1 15 - 16 1186 84
          XM_017016735.1 XP_016872224.1 15 - 16 1191 84
          XM_017016736.1 XP_016872225.1 16 - 17 1329 84
          XM_017016737.1 XP_016872226.1 14 - 15 1129 84
          XM_017016738.1 XP_016872227.1 16 - 17 1189 84
          XM_017016739.1 XP_016872228.1 17 - 18 1455 84
          GenBank mRNA AK095576.1 - 15 - 16 1275 84
          AK131199.1 - 15 - 16 1159 84
          AK225771.1 - 16 - 17 1299 84
          AK291704.1 - 15 - 16 1228 84
          AK301180.1 - 8 - 9 629 84
          BC021269.2 - 13 - 14 911 84
          BC034044.1 - 16 - 17 1254 4813948 84
          L07044.1 - 15 - 16 1127 84
          U32473.1 - 2 - 3 131 84
          U66063.1 - 5 - 6 281 84

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CAMK2G

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          MAPK cascade
          protein dephosphorylation
          calcium ion transport
          nervous system development
          regulation of skeletal muscle adaptation
          insulin secretion
          cell differentiation
          positive regulation of GTPase activity
          protein autophosphorylation
          protein complex oligomerization
          regulation of calcium ion transport
          interferon-gamma-mediated signaling pathway
          regulation of cellular response to heat
          regulation of relaxation of cardiac muscle
          protein serine/threonine kinase activity
          calmodulin-dependent protein kinase activity
          calcium-dependent protein serine/threonine phosphatase activity
          protein binding
          calmodulin binding
          ATP binding
          protein homodimerization activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0