Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00974750_m1
          See other UNC79 GE Assays ›
          Gene Symbol:
          UNC79
          Gene Name:
          unc-79 homolog (C. elegans)
          Gene Aliases:
          KIAA1409
          Chromosome Location:
          Chr.14: 93333219 - 93710473 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00974750_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: UNC79
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020818.3 NP_065869.3 38 - 39 6526 105
          XM_011537018.2 XP_011535320.1 40 - 41 9159 105
          XM_011537020.2 XP_011535322.1 39 - 40 9093 105
          XM_011537021.2 XP_011535323.1 40 - 41 6710 105
          XM_011537022.2 XP_011535324.1 40 - 41 9003 105
          XM_011537023.2 XP_011535325.1 40 - 41 9000 105
          XM_011537024.2 XP_011535326.1 39 - 40 8937 105
          XM_011537025.2 XP_011535327.1 40 - 41 9159 105
          XM_011537026.2 XP_011535328.1 40 - 41 9652 105
          XM_011537027.2 XP_011535329.1 40 - 41 6677 105
          XM_011537028.2 XP_011535330.1 39 - 40 6676 105
          XM_011537029.2 XP_011535331.1 31 - 32 5625 105
          XM_017021506.1 XP_016876995.1 40 - 41 9057 105
          XM_017021507.1 XP_016876996.1 39 - 40 9042 105
          XM_017021508.1 XP_016876997.1 39 - 40 9009 105
          XM_017021509.1 XP_016876998.1 37 - 38 9000 105
          XM_017021510.1 XP_016876999.1 38 - 39 8976 105
          XM_017021511.1 XP_016877000.1 35 - 36 5953 105
          XM_017021512.1 XP_016877001.1 39 - 40 6644 105
          XM_017021513.1 XP_016877002.1 38 - 39 8943 105
          XM_017021514.1 XP_016877003.1 38 - 39 8892 105
          XM_017021515.1 XP_016877004.1 40 - 41 6558 105
          XM_017021516.1 XP_016877005.1 37 - 38 8826 105
          XM_017021517.1 XP_016877006.1 37 - 38 8667 105
          XM_017021518.1 XP_016877007.1 34 - 35 7081 105
          XM_017021519.1 XP_016877008.1 30 - 31 6175 105
          XM_017021520.1 XP_016877009.1 24 - 25 4480 105
          GenBank mRNA AB037830.2 - 36 - 37 5880 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400008034.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14000577.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene UNC79

          Gene Ontology Categories:

          Function(s)

          ion transmembrane transport
          multicellular organism growth
          behavioral response to ethanol

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR14007101.hg.1 ENST00000256339.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14007101.hg.1 ENST00000393151.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14007101.hg.1 ENST00000393153.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14007101.hg.1 ENST00000553484.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14007101.hg.1 ENST00000555664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14007101.hg.1 NM_020818.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14007101.hg.1 TC14000577.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1400156948.hg.1 ENST00000256339.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1400156948.hg.1 ENST00000393151.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400156948.hg.1 ENST00000553484.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400156948.hg.1 ENST00000555664.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400156948.hg.1 ENST00000621021.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400156948.hg.1 NM_020818.4 Refseq
          Affymetrix Human Clariom D Assay PSR1400156948.hg.1 TC1400008034.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400156948.hg.1 ENST00000553484.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400156948.hg.1 ENST00000256339.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1400156948.hg.1 ENST00000393151.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400156948.hg.1 ENST00000555664.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400156948.hg.1 ENST00000621021.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400156948.hg.1 NM_020818.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0