Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009922.hg.1 |
ENST00000233957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009922.hg.1 |
ENST00000409599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009922.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009922.hg.1 |
TC02004995.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000334376.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000334376.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000334376.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000334376.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000466357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000466357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000466357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000466357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
ENST00000466357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009923.hg.1 |
ENST00000233957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009923.hg.1 |
ENST00000409599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009923.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009923.hg.1 |
TC02004995.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0200222876.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222876.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222876.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222876.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222876.hg.1 |
TC0200016502.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200222876.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222876.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222876.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222876.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000334376.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000334376.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000334376.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000334376.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000466357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000466357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000466357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000466357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
ENST00000466357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016502.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222877.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222877.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222877.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222877.hg.1 |
NM_003855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222877.hg.1 |
TC0200016502.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200222877.hg.1 |
ENST00000233957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222877.hg.1 |
ENST00000409599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222877.hg.1 |
NM_001282399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222877.hg.1 |
NM_003855.3 |
Refseq |