Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs00986611_m1
See other ANK1 GE Assays ›
Gene Symbol:
ANK1
Gene Name:
ankyrin 1
Gene Aliases:
ANK, SPH1, SPH2
Chromosome Location:
Chr.8: 41653225 - 41896762 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs00986611_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ANK1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_000037.3
NP_000028.3
40 - 41
5565
69
NM_001142445.1
NP_001135917.1
2 - 3
569
69
NM_001142446.1
NP_001135918.1
41 - 42
5886
69
NM_020475.2
NP_065208.2
40 - 41
5565
69
NM_020476.2
NP_065209.2
40 - 41
5565
69
NM_020477.2
NP_065210.2
40 - 41
5079
69
NM_020478.4
NP_065211.2
2 - 3
569
69
XM_005273476.4
XP_005273533.1
41 - 42
5863
69
XM_011544490.2
XP_011542792.1
43 - 44
5830
69
XM_011544491.2
XP_011542793.1
43 - 44
5829
69
XM_011544494.2
XP_011542796.1
42 - 43
5784
69
XM_011544495.2
XP_011542797.1
42 - 43
5783
69
XM_011544496.2
XP_011542798.1
43 - 44
5830
69
XM_011544500.2
XP_011542802.1
40 - 41
5667
69
XM_011544501.2
XP_011542803.1
40 - 41
5667
69
XM_011544502.2
XP_011542804.1
40 - 41
5667
69
XM_011544503.2
XP_011542805.1
42 - 43
5296
69
XM_011544504.2
XP_011542806.1
40 - 41
5181
69
XM_011544505.2
XP_011542807.1
40 - 41
5181
69
XM_017013319.1
XP_016868808.1
42 - 43
5805
69
XM_017013321.1
XP_016868810.1
43 - 44
5774
69
XM_017013322.1
XP_016868811.1
42 - 43
5736
69
XM_017013323.1
XP_016868812.1
43 - 44
5750
69
XM_017013324.1
XP_016868813.1
41 - 42
5863
69
XM_017013325.1
XP_016868814.1
42 - 43
5675
69
XM_017013326.1
XP_016868815.1
41 - 42
5623
69
XM_017013327.1
XP_016868816.1
43 - 44
5343
69
XM_017013328.1
XP_016868817.1
42 - 43
5298
69
XM_017013329.1
XP_016868818.1
41 - 42
5375
69
GenBank mRNA
AB209418.1
-
41 - 42
5886
69
AK126647.1
-
24 - 25
2959
69
AK129864.1
-
2 - 3
522
69
AK223578.1
-
2 - 3
490
69
BC007930.2
-
2 - 3
544
69
BC030957.1
-
2 - 3
557
69
BX647936.1
-
18 - 19
2878
69
M28880.1
-
40 - 41
5565
69
X16609.1
-
40 - 41
5565
69
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length less than or equal to 70
3' Most
Mammalian Gene Collection
Recommended Assay
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene ANK1
Gene Ontology Categories:
Function(s)
Process(es)
exocytosis
endoplasmic reticulum to Golgi vesicle-mediated transport
cytoskeleton organization
signal transduction
positive regulation of organelle organization
maintenance of epithelial cell apical/basal polarity
protein localization to plasma membrane
structural molecule activity
structural constituent of cytoskeleton
protein binding
cytoskeletal anchor activity
enzyme binding
spectrin binding
ATPase binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-867f8cf58d-sfwzd:80/100.66.130.62:80.
git-commit: 7a7e1ef32845d1da800a461bcf18a29ea3e3afc0
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix