Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005136.hg.1 |
TC20000358.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005136.hg.1 |
ENST00000342644.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005136.hg.1 |
ENST00000344780.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005136.hg.1 |
ENST00000372541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005136.hg.1 |
ENST00000372542.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005136.hg.1 |
ENST00000372545.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005136.hg.1 |
ENST00000462307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005136.hg.1 |
ENST00000491381.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005136.hg.1 |
NM_001042632.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005136.hg.1 |
NM_001042633.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005136.hg.1 |
NM_033421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005136.hg.1 |
NM_152897.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000342644.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000344780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000372541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000372542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000372547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000462307.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000465997.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000472219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000478230.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000486336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000491381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000614929.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
NM_001042632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
NM_001042633.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
NM_033421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
NM_152897.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
TC2000007504.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000372541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000342644.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000344780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000372542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000372547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000462307.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000465997.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000472219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000478230.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000486336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000491381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
ENST00000614929.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
NM_001042632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
NM_001042633.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
NM_033421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152297.hg.1 |
NM_152897.2 |
Refseq |