Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00990732_m1
          See other CD34 GE Assays ›
          Gene Symbol:
          CD34
          Gene Name:
          CD34 molecule
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 207886538 - 207911338 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00990732_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CD34
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025109.1 NP_001020280.1 3 - 4 778 91
          NM_001773.2 NP_001764.1 3 - 4 778 91
          GenBank mRNA AF523361.1 - 3 - 4 591 91
          AF538601.1 - 2 - 3 429 91
          AK291529.1 - 3 - 4 531 91
          AK293030.1 - 3 - 4 555 91
          AK300835.1 - 4 - 5 595 91
          BC039146.1 - 3 - 4 528 4746591 91
          M81104.1 - 3 - 4 777 91
          S53910.1 - 3 - 4 610 91
          S53911.1 - 3 - 4 610 91

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Human Hematopoeisis 1700 Inventoried Human_Hematopoisis Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CD34

          Panther Classification:

          Molecular Function -

          cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          tissue homeostasis
          endothelial cell proliferation
          glomerular filtration
          endothelium development
          cell-matrix adhesion
          signal transduction
          regulation of blood pressure
          cell proliferation
          positive regulation of gene expression
          negative regulation of gene expression
          single organismal cell-cell adhesion
          hemopoiesis
          negative regulation of blood coagulation
          negative regulation of tumor necrosis factor production
          positive regulation of interleukin-10 production
          mesangial cell-matrix adhesion
          paracrine signaling
          positive regulation of odontogenesis
          negative regulation of nitric oxide biosynthetic process
          positive regulation of angiogenesis
          cell motility
          regulation of immune response
          leukocyte migration
          transdifferentiation
          vascular wound healing
          hematopoietic stem cell proliferation
          positive regulation of transforming growth factor beta production
          positive regulation of granulocyte colony-stimulating factor production
          extracellular exosome assembly
          glomerular endothelium development
          stem cell proliferation
          metanephric glomerular mesangial cell differentiation
          negative regulation of cellular response to heat
          negative regulation of cellular response to hypoxia
          negative regulation of interleukin-2 secretion
          positive regulation of glial cell-derived neurotrophic factor secretion
          negative regulation of neuron death
          positive regulation of vasculogenesis
          transcription factor binding
          carbohydrate binding
          sulfate binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 213612 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0