Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001691.hg.1 |
ENST00000302196.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001691.hg.1 |
ENST00000378993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001691.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001691.hg.1 |
TC0X000133.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
ENST00000302196.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
ENST00000302196.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
ENST00000302196.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
ENST00000302196.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
ENST00000302196.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146898.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146898.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146898.hg.1 |
TC0X00006865.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00146898.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146898.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
ENST00000302196.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
ENST00000302196.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
ENST00000302196.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
ENST00000302196.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
ENST00000302196.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
ENST00000378993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006865.hg.1 |
NM_014271.3 |
Refseq |