Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006937.hg.1 |
ENST00000315323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006937.hg.1 |
ENST00000541149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006937.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006937.hg.1 |
TC17000571.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006935.hg.1 |
ENST00000541149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006935.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006935.hg.1 |
TC17000571.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006936.hg.1 |
ENST00000315323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006936.hg.1 |
ENST00000541149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006936.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006936.hg.1 |
TC17000571.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008005.hg.1 |
ENST00000315323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008005.hg.1 |
ENST00000315323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008005.hg.1 |
ENST00000315323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008005.hg.1 |
ENST00000315323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008005.hg.1 |
ENST00000315323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008005.hg.1 |
ENST00000315323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008005.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008005.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008005.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008005.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008005.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008005.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008005.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006939.hg.1 |
ENST00000315323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006939.hg.1 |
ENST00000541149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006939.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006939.hg.1 |
TC17000571.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006938.hg.1 |
ENST00000315323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006938.hg.1 |
ENST00000541149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006938.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006938.hg.1 |
TC17000571.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700158580.hg.1 |
ENST00000315323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700158580.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700158580.hg.1 |
TC1700008005.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700158580.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700158580.hg.1 |
ENST00000315323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008005.hg.1 |
ENST00000315323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008005.hg.1 |
ENST00000315323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008005.hg.1 |
ENST00000315323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008005.hg.1 |
ENST00000315323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008005.hg.1 |
ENST00000315323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008005.hg.1 |
ENST00000315323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008005.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008005.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008005.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008005.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008005.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008005.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008005.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700158578.hg.1 |
ENST00000315323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700158578.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700158578.hg.1 |
TC1700008005.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700158578.hg.1 |
ENST00000315323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700158578.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700158582.hg.1 |
ENST00000315323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700158582.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700158582.hg.1 |
TC1700008005.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700158582.hg.1 |
ENST00000315323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700158582.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700158579.hg.1 |
ENST00000315323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700158579.hg.1 |
NM_001466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700158579.hg.1 |
TC1700008005.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700158579.hg.1 |
ENST00000315323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700158579.hg.1 |
NM_001466.3 |
Refseq |