Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02047649.hg.1 |
ENST00000305784.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02047649.hg.1 |
NM_001165031.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02047649.hg.1 |
NM_012145.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02047649.hg.1 |
NR_033255.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02047649.hg.1 |
TC02002954.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0200220241.hg.1 |
ENST00000305784.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200220241.hg.1 |
ENST00000400770.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200220241.hg.1 |
ENST00000420144.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200220241.hg.1 |
ENST00000432348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200220241.hg.1 |
ENST00000445261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200220241.hg.1 |
ENST00000464603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200220241.hg.1 |
NM_001165031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200220241.hg.1 |
NM_012145.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200220241.hg.1 |
NR_033255.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200220241.hg.1 |
TC0200016369.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200220241.hg.1 |
ENST00000305784.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200220241.hg.1 |
ENST00000400770.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200220241.hg.1 |
ENST00000420144.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200220241.hg.1 |
ENST00000432348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200220241.hg.1 |
ENST00000445261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200220241.hg.1 |
ENST00000464603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200220241.hg.1 |
NM_001165031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200220241.hg.1 |
NM_012145.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200220241.hg.1 |
NR_033255.1 |
Refseq |