Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008194.hg.1 |
TC22000433.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008194.hg.1 |
ENST00000008876.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008194.hg.1 |
ENST00000329492.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008194.hg.1 |
ENST00000341339.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008194.hg.1 |
ENST00000399908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008194.hg.1 |
ENST00000399912.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008194.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008197.hg.1 |
TC22000433.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008197.hg.1 |
ENST00000008876.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008197.hg.1 |
ENST00000329492.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008197.hg.1 |
ENST00000341339.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008197.hg.1 |
ENST00000399908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008197.hg.1 |
ENST00000399912.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008197.hg.1 |
ENST00000442429.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008197.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008191.hg.1 |
TC22000433.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008191.hg.1 |
ENST00000008876.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008191.hg.1 |
ENST00000329492.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008191.hg.1 |
ENST00000399908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008191.hg.1 |
ENST00000399912.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008191.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008193.hg.1 |
TC22000433.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008193.hg.1 |
ENST00000008876.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008193.hg.1 |
ENST00000329492.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008193.hg.1 |
ENST00000399908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008193.hg.1 |
ENST00000399912.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008193.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008195.hg.1 |
TC22000433.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008195.hg.1 |
ENST00000008876.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008195.hg.1 |
ENST00000329492.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008195.hg.1 |
ENST00000341339.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008195.hg.1 |
ENST00000399908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008195.hg.1 |
ENST00000399912.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008195.hg.1 |
ENST00000442429.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008195.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008192.hg.1 |
TC22000433.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008192.hg.1 |
ENST00000329492.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008192.hg.1 |
ENST00000399908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008192.hg.1 |
ENST00000399912.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008192.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169572.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169572.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169572.hg.1 |
TC2200009303.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200169572.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169572.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169573.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169573.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169573.hg.1 |
TC2200009303.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200169573.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169573.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
ENST00000329492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009303.hg.1 |
NM_012324.5 |
Refseq |