Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04006649.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04006649.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04006649.hg.1 |
TC04000462.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
ENST00000511298.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
ENST00000511298.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
ENST00000511298.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
ENST00000511298.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
ENST00000511298.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
ENST00000511298.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400155175.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400155175.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400155175.hg.1 |
TC0400008020.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400155175.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400155175.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
ENST00000295887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
ENST00000511298.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
ENST00000511298.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
ENST00000511298.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
ENST00000511298.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
ENST00000511298.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
ENST00000511298.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008020.hg.1 |
NM_001263.3 |
Refseq |