Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
NM_001304725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
NM_001304725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
NM_001304725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
NM_001304725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
NM_001304725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
NM_001304725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
NM_001304725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
NM_001304725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018239.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012635.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012635.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012635.hg.1 |
TC01006272.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012634.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012634.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012634.hg.1 |
TC01006272.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
NM_001304725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
NM_001304725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
NM_001304725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
NM_001304725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
NM_001304725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
NM_001304725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
NM_001304725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
NM_001304725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018239.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238235.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238235.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238235.hg.1 |
TC0100018239.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100238235.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238235.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238236.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238236.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238236.hg.1 |
TC0100018239.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100238236.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238236.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238237.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238237.hg.1 |
NM_001304725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238237.hg.1 |
NM_017655.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238237.hg.1 |
TC0100018239.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100238237.hg.1 |
ENST00000370759.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238237.hg.1 |
NM_001304725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238237.hg.1 |
NM_017655.5 |
Refseq |